Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23413932-23414161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4552591 c.5647G>A p.E1883K Chr.14: 23414015-23414015 on Build GRCh38
3495201 c.5563G>A p.E1855K Chr.14: 23414099-23414099 on Build GRCh38
5514288 c.5639G>A p.R1880H Chr.14: 23414023-23414023 on Build GRCh38
4912249 c.5654C>T p.A1885V Chr.14: 23414008-23414008 on Build GRCh38
6218345 c.5602G>A p.V1868I Chr.14: 23414060-23414060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs121913652 Chr.14: 23414015-23414015 on Build GRCh38
rs749520200 Chr.14: 23414124-23414124 on Build GRCh38
rs745984048 Chr.14: 23414129-23414129 on Build GRCh38
rs797044598 Chr.14: 23414096-23414096 on Build GRCh38
rs751425608 Chr.14: 23414129-23414130 on Build GRCh38
rs767956231 Chr.14: 23414108-23414108 on Build GRCh38
rs560939959 Chr.14: 23414060-23414060 on Build GRCh38
rs903422461 Chr.14: 23414005-23414005 on Build GRCh38
rs749923218 Chr.14: 23413968-23413968 on Build GRCh38
rs368706722 Chr.14: 23414100-23414100 on Build GRCh38
rs730880824 Chr.14: 23414056-23414056 on Build GRCh38
rs761512935 Chr.14: 23413957-23413957 on Build GRCh38
rs529784690 Chr.14: 23414088-23414088 on Build GRCh38
rs753392652 Chr.14: 23414007-23414007 on Build GRCh38
rs3729499 Chr.14: 23413986-23413986 on Build GRCh38
rs886038830 Chr.14: 23414038-23414038 on Build GRCh38
rs375621171 Chr.14: 23414116-23414116 on Build GRCh38
rs730880726 Chr.14: 23414016-23414016 on Build GRCh38
rs867477685 Chr.14: 23414023-23414023 on Build GRCh38
rs778008286 Chr.14: 23414028-23414028 on Build GRCh38
rs755230416 Chr.14: 23413994-23413994 on Build GRCh38
rs896043528 Chr.14: 23413958-23413958 on Build GRCh38
rs915005545 Chr.14: 23413988-23413988 on Build GRCh38
rs749320297 Chr.14: 23414043-23414043 on Build GRCh38
rs768534661 Chr.14: 23414073-23414073 on Build GRCh38
rs112423919 Chr.14: 23414131-23414131 on Build GRCh38
rs369182253 Chr.14: 23414135-23414135 on Build GRCh38
rs780657149 Chr.14: 23413985-23413985 on Build GRCh38
rs779038045 Chr.14: 23414128-23414128 on Build GRCh38
rs778224065 Chr.14: 23414109-23414109 on Build GRCh38
rs376668612 Chr.14: 23414075-23414075 on Build GRCh38
rs367580423 Chr.14: 23414025-23414025 on Build GRCh38
rs762494719 Chr.14: 23414097-23414097 on Build GRCh38
rs3729833 Chr.14: 23413975-23413975 on Build GRCh38
rs746962437 Chr.14: 23414070-23414070 on Build GRCh38
rs762612534 Chr.14: 23414104-23414105 on Build GRCh38
rs373707937 Chr.14: 23413998-23413998 on Build GRCh38
rs765033010 Chr.14: 23413961-23413961 on Build GRCh38
rs587781086 Chr.14: 23414114-23414114 on Build GRCh38
rs755493069 Chr.14: 23414106-23414106 on Build GRCh38
rs754553717 Chr.14: 23414135-23414136 on Build GRCh38
rs766359220 Chr.14: 23414125-23414126 on Build GRCh38
rs372381770 Chr.14: 23414101-23414101 on Build GRCh38
rs45520836 Chr.14: 23414074-23414074 on Build GRCh38
rs768609796 Chr.14: 23414134-23414134 on Build GRCh38

View Full Product Details