Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113231101-113231362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

SNP Detail

SNP ID Chromosome Location
rs186090080 Chr.11: 113231305-113231305 on Build GRCh38
rs886347579 Chr.11: 113231300-113231300 on Build GRCh38
rs567243503 Chr.11: 113231335-113231335 on Build GRCh38
rs553533062 Chr.11: 113231325-113231325 on Build GRCh38
rs1016136102 Chr.11: 113231180-113231180 on Build GRCh38
rs990533101 Chr.11: 113231340-113231340 on Build GRCh38
rs782674037 Chr.11: 113231238-113231238 on Build GRCh38
rs949087630 Chr.11: 113231204-113231204 on Build GRCh38
rs782264232 Chr.11: 113231129-113231129 on Build GRCh38
rs781968088 Chr.11: 113231278-113231279 on Build GRCh38
rs781992571 Chr.11: 113231211-113231211 on Build GRCh38
rs117219783 Chr.11: 113231236-113231236 on Build GRCh38
rs1039104842 Chr.11: 113231245-113231245 on Build GRCh38
rs561631434 Chr.11: 113231172-113231172 on Build GRCh38
rs143883407 Chr.11: 113231215-113231215 on Build GRCh38
rs782724000 Chr.11: 113231246-113231246 on Build GRCh38
rs990585263 Chr.11: 113231190-113231190 on Build GRCh38
rs781831035 Chr.11: 113231251-113231251 on Build GRCh38
rs997525838 Chr.11: 113231250-113231250 on Build GRCh38
rs963103669 Chr.11: 113231189-113231189 on Build GRCh38
rs1053203333 Chr.11: 113231224-113231224 on Build GRCh38
rs549784673 Chr.11: 113231241-113231241 on Build GRCh38
rs782519655 Chr.11: 113231281-113231282 on Build GRCh38
rs530375301 Chr.11: 113231175-113231175 on Build GRCh38
rs782647733 Chr.11: 113231258-113231258 on Build GRCh38
rs963219157 Chr.11: 113231328-113231328 on Build GRCh38
rs149464405 Chr.11: 113231210-113231210 on Build GRCh38
rs554626814 Chr.11: 113231165-113231165 on Build GRCh38
rs565218399 Chr.11: 113231312-113231312 on Build GRCh38
rs982060994 Chr.11: 113231207-113231207 on Build GRCh38
rs374188281 Chr.11: 113231254-113231254 on Build GRCh38
rs267602694 Chr.11: 113231235-113231235 on Build GRCh38
rs916219634 Chr.11: 113231203-113231203 on Build GRCh38
rs983299268 Chr.11: 113231177-113231177 on Build GRCh38
rs782539999 Chr.11: 113231308-113231308 on Build GRCh38

View Full Product Details