Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93741335-93741606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
UniGene
Hs.600086
Gene Chromosome Location
Chr.5: 93615130-94111699 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543671.2 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NM_153216.1

Gene Symbol
POU5F2
Entrez Gene ID
134187
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
SPRM-1
UniGene
Hs.678995
Gene Chromosome Location
Chr.5: 93740309-93741603 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543671.2 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NM_153216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3856909 c.103T>G p.L35V Chr.5: 93741461-93741461 on Build GRCh38
3828604 c.183G>C p.W61C Chr.5: 93741381-93741381 on Build GRCh38
4169464 c.179T>G p.V60G Chr.5: 93741385-93741385 on Build GRCh38
6171750 c.178G>A p.V60M Chr.5: 93741386-93741386 on Build GRCh38
3982079 c.203T>G p.L68R Chr.5: 93741361-93741361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368188217 Chr.5: 93741410-93741410 on Build GRCh38
rs112317301 Chr.5: 93741584-93741584 on Build GRCh38
rs759366415 Chr.5: 93741360-93741360 on Build GRCh38
rs767125228 Chr.5: 93741558-93741558 on Build GRCh38
rs993920752 Chr.5: 93741559-93741559 on Build GRCh38
rs753112880 Chr.5: 93741397-93741397 on Build GRCh38
rs760753919 Chr.5: 93741386-93741386 on Build GRCh38
rs138308146 Chr.5: 93741387-93741387 on Build GRCh38
rs745568308 Chr.5: 93741429-93741429 on Build GRCh38
rs376581154 Chr.5: 93741456-93741456 on Build GRCh38
rs754087969 Chr.5: 93741390-93741390 on Build GRCh38
rs752849590 Chr.5: 93741557-93741558 on Build GRCh38
rs776757072 Chr.5: 93741530-93741530 on Build GRCh38
rs779590395 Chr.5: 93741512-93741512 on Build GRCh38
rs754742643 Chr.5: 93741580-93741580 on Build GRCh38
rs372897251 Chr.5: 93741417-93741417 on Build GRCh38
rs775262614 Chr.5: 93741469-93741469 on Build GRCh38
rs762990553 Chr.5: 93741367-93741367 on Build GRCh38
rs770598240 Chr.5: 93741423-93741423 on Build GRCh38
rs780945503 Chr.5: 93741427-93741427 on Build GRCh38
rs181777565 Chr.5: 93741436-93741436 on Build GRCh38
rs557446896 Chr.5: 93741546-93741546 on Build GRCh38
rs760553819 Chr.5: 93741484-93741484 on Build GRCh38
rs768577139 Chr.5: 93741437-93741437 on Build GRCh38
rs372798416 Chr.5: 93741528-93741528 on Build GRCh38
rs555270643 Chr.5: 93741448-93741448 on Build GRCh38
rs774222267 Chr.5: 93741519-93741519 on Build GRCh38
rs769448417 Chr.5: 93741435-93741435 on Build GRCh38
rs374687071 Chr.5: 93741405-93741405 on Build GRCh38
rs760702752 Chr.5: 93741443-93741443 on Build GRCh38
rs766466059 Chr.5: 93741444-93741444 on Build GRCh38
rs963985123 Chr.5: 93741459-93741459 on Build GRCh38
rs977984699 Chr.5: 93741529-93741529 on Build GRCh38
rs748131448 Chr.5: 93741521-93741521 on Build GRCh38
rs575553090 Chr.5: 93741506-93741506 on Build GRCh38
rs753162634 Chr.5: 93741481-93741481 on Build GRCh38
rs755580301 Chr.5: 93741572-93741572 on Build GRCh38
rs748912839 Chr.5: 93741579-93741579 on Build GRCh38
rs538047369 Chr.5: 93741542-93741542 on Build GRCh38
rs994314747 Chr.5: 93741402-93741408 on Build GRCh38
rs768364996 Chr.5: 93741515-93741515 on Build GRCh38
rs767585902 Chr.5: 93741421-93741421 on Build GRCh38
rs1026226878 Chr.5: 93741407-93741407 on Build GRCh38
rs755856638 Chr.5: 93741507-93741507 on Build GRCh38
rs765432822 Chr.5: 93741534-93741534 on Build GRCh38
rs577438749 Chr.5: 93741550-93741550 on Build GRCh38
rs749040900 Chr.5: 93741513-93741513 on Build GRCh38
rs749895032 Chr.5: 93741563-93741563 on Build GRCh38
rs371634969 Chr.5: 93741447-93741447 on Build GRCh38
rs767493941 Chr.5: 93741372-93741372 on Build GRCh38
rs369781509 Chr.5: 93741504-93741504 on Build GRCh38
rs372081385 Chr.5: 93741395-93741395 on Build GRCh38
rs775270280 Chr.5: 93741362-93741362 on Build GRCh38
rs947665086 Chr.5: 93741574-93741574 on Build GRCh38
rs769654563 Chr.5: 93741361-93741361 on Build GRCh38
rs1040792454 Chr.5: 93741518-93741518 on Build GRCh38
rs755246968 Chr.5: 93741391-93741391 on Build GRCh38
rs752135459 Chr.5: 93741496-93741496 on Build GRCh38
rs375465714 Chr.5: 93741585-93741585 on Build GRCh38
rs774272055 Chr.5: 93741438-93741438 on Build GRCh38
rs758694909 Chr.5: 93741493-93741493 on Build GRCh38
rs757909429 Chr.5: 93741552-93741552 on Build GRCh38
rs370809672 Chr.5: 93741561-93741561 on Build GRCh38
rs373831731 Chr.5: 93741541-93741541 on Build GRCh38
rs779713081 Chr.5: 93741577-93741577 on Build GRCh38
rs780892278 Chr.5: 93741505-93741505 on Build GRCh38

View Full Product Details