Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27623142-27623367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1D
Entrez Gene ID
51082
Gene Name
RNA polymerase I subunit D
Gene Aliases
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2
UniGene
Hs.507584
Gene Chromosome Location
Chr.13: 27620743-27667422 on Build GRCh38
Transcripts
NM_001206559.1 NM_015972.3 NM_152705.2 XM_005266412.1 XM_005266414.2 XM_017020623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793229 c.320G>A p.C107Y Chr.13: 27623168-27623168 on Build GRCh38
6564481 c.300G>A p.L100L Chr.13: 27623148-27623148 on Build GRCh38
7222101 c.334G>C p.D112H Chr.13: 27623182-27623182 on Build GRCh38
4716512 c.346G>A p.A116T Chr.13: 27623194-27623194 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886050109 Chr.13: 27623207-27623207 on Build GRCh38
rs777942743 Chr.13: 27623210-27623210 on Build GRCh38
rs376204257 Chr.13: 27623271-27623271 on Build GRCh38
rs760229520 Chr.13: 27623173-27623173 on Build GRCh38
rs779011956 Chr.13: 27623261-27623261 on Build GRCh38
rs921126613 Chr.13: 27623220-27623220 on Build GRCh38
rs765667059 Chr.13: 27623196-27623196 on Build GRCh38
rs747909163 Chr.13: 27623264-27623264 on Build GRCh38
rs534978633 Chr.13: 27623228-27623228 on Build GRCh38
rs8459 Chr.13: 27623280-27623280 on Build GRCh38
rs770341695 Chr.13: 27623293-27623293 on Build GRCh38
rs1014318222 Chr.13: 27623341-27623341 on Build GRCh38
rs751829978 Chr.13: 27623218-27623218 on Build GRCh38
rs771862796 Chr.13: 27623285-27623285 on Build GRCh38
rs772029302 Chr.13: 27623266-27623266 on Build GRCh38
rs553357664 Chr.13: 27623343-27623343 on Build GRCh38
rs7097 Chr.13: 27623299-27623299 on Build GRCh38
rs954802306 Chr.13: 27623305-27623305 on Build GRCh38
rs758795471 Chr.13: 27623202-27623202 on Build GRCh38
rs781403620 Chr.13: 27623231-27623231 on Build GRCh38
rs745943558 Chr.13: 27623255-27623255 on Build GRCh38
rs772974457 Chr.13: 27623278-27623278 on Build GRCh38
rs778936453 Chr.13: 27623247-27623247 on Build GRCh38
rs932486325 Chr.13: 27623221-27623221 on Build GRCh38
rs768585316 Chr.13: 27623256-27623256 on Build GRCh38
rs987734200 Chr.13: 27623315-27623315 on Build GRCh38
rs974711550 Chr.13: 27623296-27623296 on Build GRCh38
rs771797869 Chr.13: 27623172-27623172 on Build GRCh38
rs149222191 Chr.13: 27623182-27623182 on Build GRCh38
rs934624616 Chr.13: 27623260-27623260 on Build GRCh38
rs377050034 Chr.13: 27623187-27623187 on Build GRCh38
rs886050110 Chr.13: 27623341-27623341 on Build GRCh38
rs143410332 Chr.13: 27623197-27623197 on Build GRCh38
rs138256149 Chr.13: 27623177-27623177 on Build GRCh38
rs1005108489 Chr.13: 27623194-27623194 on Build GRCh38

View Full Product Details