Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159992937-159993200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
UniGene
Hs.673
Gene Chromosome Location
Chr.3: 159988836-159996019 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Gene Symbol
IL12A-AS1
Entrez Gene ID
101928376
Gene Name
IL12A antisense RNA 1
Gene Aliases
-
UniGene
Hs.565865
Gene Chromosome Location
Chr.3: 159913400-160207092 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078933 c.331G>A p.D111N Chr.3: 159993078-159993078 on Build GRCh38
5591320 c.324C>T p.I108I Chr.3: 159993071-159993071 on Build GRCh38
3673879 c.327A>G p.T109T Chr.3: 159993074-159993074 on Build GRCh38
7142225 c.313C>A p.H105N Chr.3: 159993060-159993060 on Build GRCh38
1693693 c.346G>A p.V116M Chr.3: 159993093-159993093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768505894 Chr.3: 159992985-159992985 on Build GRCh38
rs151148465 Chr.3: 159993088-159993088 on Build GRCh38
rs772865699 Chr.3: 159993130-159993130 on Build GRCh38
rs761280149 Chr.3: 159992999-159992999 on Build GRCh38
rs182240590 Chr.3: 159993167-159993167 on Build GRCh38
rs142035807 Chr.3: 159993073-159993073 on Build GRCh38
rs1052861130 Chr.3: 159993098-159993098 on Build GRCh38
rs766795077 Chr.3: 159993021-159993021 on Build GRCh38
rs746817892 Chr.3: 159993101-159993101 on Build GRCh38
rs140235653 Chr.3: 159993032-159993032 on Build GRCh38
rs146682745 Chr.3: 159992960-159992960 on Build GRCh38
rs775481686 Chr.3: 159993110-159993110 on Build GRCh38
rs767956212 Chr.3: 159993058-159993058 on Build GRCh38
rs757491719 Chr.3: 159993095-159993095 on Build GRCh38
rs747689322 Chr.3: 159993111-159993111 on Build GRCh38
rs74321485 Chr.3: 159993053-159993053 on Build GRCh38
rs531622172 Chr.3: 159993015-159993015 on Build GRCh38
rs371082765 Chr.3: 159993170-159993171 on Build GRCh38
rs746914870 Chr.3: 159992976-159992976 on Build GRCh38
rs202231418 Chr.3: 159992969-159992969 on Build GRCh38
rs770741319 Chr.3: 159993139-159993139 on Build GRCh38
rs752804893 Chr.3: 159993061-159993061 on Build GRCh38
rs777830138 Chr.3: 159993090-159993090 on Build GRCh38
rs754001872 Chr.3: 159993092-159993092 on Build GRCh38
rs901355334 Chr.3: 159993006-159993006 on Build GRCh38
rs774754016 Chr.3: 159993145-159993145 on Build GRCh38
rs776177489 Chr.3: 159992991-159992991 on Build GRCh38
rs971399492 Chr.3: 159993040-159993040 on Build GRCh38
rs764662309 Chr.3: 159993135-159993136 on Build GRCh38
rs769557811 Chr.3: 159993010-159993010 on Build GRCh38
rs762649502 Chr.3: 159993135-159993135 on Build GRCh38
rs779783410 Chr.3: 159993097-159993097 on Build GRCh38
rs891584078 Chr.3: 159993148-159993148 on Build GRCh38
rs773066368 Chr.3: 159993013-159993013 on Build GRCh38
rs751894716 Chr.3: 159993044-159993044 on Build GRCh38
rs372252038 Chr.3: 159993121-159993121 on Build GRCh38

View Full Product Details