Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179576134-179576395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL6A
Entrez Gene ID
86
Gene Name
actin like 6A
Gene Aliases
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K
UniGene
Hs.435326
Gene Chromosome Location
Chr.3: 179562880-179588405 on Build GRCh38
Transcripts
NM_004301.4 NM_177989.3 NM_178042.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
215895 c.491G>A p.R164H Chr.3: 179576231-179576231 on Build GRCh38
4772737 c.491G>T p.R164L Chr.3: 179576231-179576231 on Build GRCh38
1041554 c.489T>G p.G163G Chr.3: 179576229-179576229 on Build GRCh38
1041555 c.490C>T p.R164C Chr.3: 179576230-179576230 on Build GRCh38
4659109 c.505A>G p.I169V Chr.3: 179576245-179576245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779678440 Chr.3: 179576205-179576205 on Build GRCh38
rs767027212 Chr.3: 179576320-179576320 on Build GRCh38
rs758112188 Chr.3: 179576204-179576204 on Build GRCh38
rs207464113 Chr.3: 179576177-179576177 on Build GRCh38
rs750572315 Chr.3: 179576344-179576345 on Build GRCh38
rs202120446 Chr.3: 179576298-179576298 on Build GRCh38
rs374025900 Chr.3: 179576351-179576351 on Build GRCh38
rs376704622 Chr.3: 179576289-179576289 on Build GRCh38
rs148839195 Chr.3: 179576178-179576178 on Build GRCh38
rs759486123 Chr.3: 179576334-179576334 on Build GRCh38
rs758999228 Chr.3: 179576307-179576307 on Build GRCh38
rs201109652 Chr.3: 179576264-179576264 on Build GRCh38
rs35449075 Chr.3: 179576241-179576241 on Build GRCh38
rs533137497 Chr.3: 179576337-179576337 on Build GRCh38
rs761828879 Chr.3: 179576168-179576170 on Build GRCh38
rs770077344 Chr.3: 179576297-179576297 on Build GRCh38
rs137876412 Chr.3: 179576283-179576283 on Build GRCh38
rs746466516 Chr.3: 179576234-179576234 on Build GRCh38
rs543773000 Chr.3: 179576354-179576354 on Build GRCh38
rs939181350 Chr.3: 179576231-179576231 on Build GRCh38
rs771113722 Chr.3: 179576306-179576306 on Build GRCh38
rs750041679 Chr.3: 179576193-179576193 on Build GRCh38
rs764989960 Chr.3: 179576179-179576179 on Build GRCh38
rs368778776 Chr.3: 179576301-179576301 on Build GRCh38
rs781498873 Chr.3: 179576271-179576271 on Build GRCh38
rs765309611 Chr.3: 179576341-179576341 on Build GRCh38

View Full Product Details