Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39025385-39025644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3D
Entrez Gene ID
140564
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3D
Gene Aliases
A3D, APOBEC3DE, APOBEC3E, ARP6
UniGene
Hs.629775
Gene Chromosome Location
Chr.22: 39021113-39034328 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Gene Symbol
APOBEC3F
Entrez Gene ID
200316
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Gene Aliases
A3F, ARP8, BK150C2.4.MRNA, KA6
UniGene
Hs.659809
Gene Chromosome Location
Chr.22: 39020454-39055972 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5484509 c.677C>A p.A226D Chr.22: 39025536-39025536 on Build GRCh38
6414621 c.505T>G p.W169G Chr.22: 39025571-39025571 on Build GRCh38
259468 c.765C>T p.F255F Chr.22: 39025624-39025624 on Build GRCh38
6162252 c.577C>A p.L193M Chr.22: 39025643-39025643 on Build GRCh38
4918968 c.777T>C p.Y259Y Chr.22: 39025636-39025636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367844222 Chr.22: 39025527-39025527 on Build GRCh38
rs114229069 Chr.22: 39025504-39025504 on Build GRCh38
rs772270455 Chr.22: 39025487-39025487 on Build GRCh38
rs764332727 Chr.22: 39025432-39025432 on Build GRCh38
rs749069463 Chr.22: 39025470-39025470 on Build GRCh38
rs143474193 Chr.22: 39025422-39025422 on Build GRCh38
rs139309648 Chr.22: 39025581-39025581 on Build GRCh38
rs752289272 Chr.22: 39025454-39025454 on Build GRCh38
rs754429373 Chr.22: 39025448-39025448 on Build GRCh38
rs200163974 Chr.22: 39025521-39025521 on Build GRCh38
rs775676692 Chr.22: 39025488-39025488 on Build GRCh38
rs766985357 Chr.22: 39025520-39025520 on Build GRCh38
rs368121641 Chr.22: 39025558-39025558 on Build GRCh38
rs201658890 Chr.22: 39025607-39025607 on Build GRCh38
rs773322348 Chr.22: 39025545-39025547 on Build GRCh38
rs770853742 Chr.22: 39025471-39025471 on Build GRCh38
rs748431888 Chr.22: 39025545-39025545 on Build GRCh38
rs528328922 Chr.22: 39025608-39025608 on Build GRCh38
rs768598048 Chr.22: 39025519-39025520 on Build GRCh38
rs115210060 Chr.22: 39025507-39025507 on Build GRCh38
rs376455314 Chr.22: 39025443-39025443 on Build GRCh38
rs959721511 Chr.22: 39025458-39025458 on Build GRCh38
rs190831871 Chr.22: 39025602-39025602 on Build GRCh38
rs770142981 Chr.22: 39025557-39025557 on Build GRCh38
rs777417359 Chr.22: 39025526-39025526 on Build GRCh38
rs1001399987 Chr.22: 39025593-39025593 on Build GRCh38
rs749341019 Chr.22: 39025519-39025519 on Build GRCh38
rs754274350 Chr.22: 39025435-39025435 on Build GRCh38
rs762174544 Chr.22: 39025508-39025508 on Build GRCh38
rs776857077 Chr.22: 39025544-39025544 on Build GRCh38
rs778934213 Chr.22: 39025481-39025481 on Build GRCh38
rs370274357 Chr.22: 39025532-39025532 on Build GRCh38
rs146744584 Chr.22: 39025512-39025512 on Build GRCh38
rs201911698 Chr.22: 39025535-39025535 on Build GRCh38
rs377283514 Chr.22: 39025485-39025485 on Build GRCh38
rs765098359 Chr.22: 39025595-39025595 on Build GRCh38
rs760172009 Chr.22: 39025585-39025585 on Build GRCh38
rs907553690 Chr.22: 39025491-39025491 on Build GRCh38
rs778917803 Chr.22: 39025529-39025529 on Build GRCh38
rs186672417 Chr.22: 39025559-39025559 on Build GRCh38
rs201373761 Chr.22: 39025587-39025587 on Build GRCh38
rs768625030 Chr.22: 39025543-39025543 on Build GRCh38
rs755602956 Chr.22: 39025523-39025523 on Build GRCh38
rs375390945 Chr.22: 39025534-39025534 on Build GRCh38
rs777408528 Chr.22: 39025462-39025462 on Build GRCh38
rs1049952294 Chr.22: 39025515-39025515 on Build GRCh38
rs747068569 Chr.22: 39025541-39025541 on Build GRCh38
rs780030323 Chr.22: 39025447-39025447 on Build GRCh38
rs763570476 Chr.22: 39025586-39025586 on Build GRCh38
rs750919260 Chr.22: 39025514-39025514 on Build GRCh38
rs963420093 Chr.22: 39025500-39025500 on Build GRCh38
rs760781145 Chr.22: 39025489-39025489 on Build GRCh38
rs745764932 Chr.22: 39025533-39025533 on Build GRCh38
rs550306184 Chr.22: 39025418-39025418 on Build GRCh38
rs766861928 Chr.22: 39025561-39025561 on Build GRCh38
rs763519936 Chr.22: 39025516-39025516 on Build GRCh38
rs765696442 Chr.22: 39025439-39025439 on Build GRCh38
rs371292817 Chr.22: 39025451-39025451 on Build GRCh38
rs374339162 Chr.22: 39025474-39025474 on Build GRCh38
rs755830934 Chr.22: 39025461-39025461 on Build GRCh38
rs200273887 Chr.22: 39025590-39025590 on Build GRCh38
rs757104391 Chr.22: 39025528-39025528 on Build GRCh38

View Full Product Details