Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 64541469-64541694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS9
Entrez Gene ID
56999
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Gene Aliases
-
UniGene
Hs.656071
Gene Chromosome Location
Chr.3: 64515654-64688000 on Build GRCh38
Transcripts
NM_001318781.1 NM_182920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
109135 c.5265C>T p.F1755F Chr.3: 64541553-64541553 on Build GRCh38
3916586 c.5292G>A p.K1764K Chr.3: 64541526-64541526 on Build GRCh38
3916587 c.5276G>A p.R1759K Chr.3: 64541542-64541542 on Build GRCh38
3373284 c.5230A>G p.R1744G Chr.3: 64541588-64541588 on Build GRCh38
4551199 c.5220G>A p.K1740K Chr.3: 64541598-64541598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748929453 Chr.3: 64541620-64541620 on Build GRCh38
rs776637203 Chr.3: 64541617-64541617 on Build GRCh38
rs372558553 Chr.3: 64541636-64541636 on Build GRCh38
rs139232267 Chr.3: 64541552-64541552 on Build GRCh38
rs368793950 Chr.3: 64541504-64541504 on Build GRCh38
rs753239746 Chr.3: 64541534-64541534 on Build GRCh38
rs1022918445 Chr.3: 64541655-64541655 on Build GRCh38
rs149034225 Chr.3: 64541553-64541553 on Build GRCh38
rs757669493 Chr.3: 64541649-64541649 on Build GRCh38
rs766188311 Chr.3: 64541616-64541616 on Build GRCh38
rs374739163 Chr.3: 64541579-64541579 on Build GRCh38
rs760571202 Chr.3: 64541514-64541514 on Build GRCh38
rs771388020 Chr.3: 64541578-64541578 on Build GRCh38
rs764417053 Chr.3: 64541638-64541638 on Build GRCh38
rs750385148 Chr.3: 64541512-64541512 on Build GRCh38
rs771965779 Chr.3: 64541601-64541601 on Build GRCh38
rs772929869 Chr.3: 64541611-64541611 on Build GRCh38
rs190978193 Chr.3: 64541511-64541511 on Build GRCh38
rs752341250 Chr.3: 64541634-64541634 on Build GRCh38
rs756262487 Chr.3: 64541661-64541661 on Build GRCh38
rs17070967 Chr.3: 64541599-64541599 on Build GRCh38
rs1036919 Chr.3: 64541660-64541660 on Build GRCh38
rs369063444 Chr.3: 64541595-64541595 on Build GRCh38
rs759116845 Chr.3: 64541550-64541550 on Build GRCh38
rs781496825 Chr.3: 64541653-64541653 on Build GRCh38
rs1373163 Chr.3: 64541645-64541645 on Build GRCh38
rs141560382 Chr.3: 64541542-64541542 on Build GRCh38
rs892975721 Chr.3: 64541647-64541647 on Build GRCh38
rs778468835 Chr.3: 64541543-64541543 on Build GRCh38
rs747515768 Chr.3: 64541563-64541563 on Build GRCh38
rs778176536 Chr.3: 64541562-64541562 on Build GRCh38
rs552717987 Chr.3: 64541612-64541612 on Build GRCh38
rs896562498 Chr.3: 64541528-64541528 on Build GRCh38
rs372386459 Chr.3: 64541521-64541521 on Build GRCh38
rs200699376 Chr.3: 64541626-64541626 on Build GRCh38
rs764784401 Chr.3: 64541631-64541631 on Build GRCh38

View Full Product Details