Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100522438-100522665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD46
Entrez Gene ID
157567
Gene Name
ankyrin repeat domain 46
Gene Aliases
ANK-S, GENX-115279
UniGene
Hs.530199
Gene Chromosome Location
Chr.8: 100509752-100559786 on Build GRCh38
Transcripts
NM_001270377.1 NM_001270378.1 NM_001270379.1 NM_198401.3 XM_006716509.1 XM_017013106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
485876 c.605T>C p.I202T Chr.8: 100522637-100522637 on Build GRCh38
748087 c.637G>A p.V213M Chr.8: 100522605-100522605 on Build GRCh38
4759483 c.662A>T p.E221V Chr.8: 100522580-100522580 on Build GRCh38
4860027 c.636+1G>A p.? Chr.8: 100522605-100522605 on Build GRCh38
485877 c.605T>C p.I202T Chr.8: 100522637-100522637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747566340 Chr.8: 100522562-100522562 on Build GRCh38
rs747335527 Chr.8: 100522513-100522513 on Build GRCh38
rs763554095 Chr.8: 100522606-100522606 on Build GRCh38
rs1044504 Chr.8: 100522524-100522524 on Build GRCh38
rs34603431 Chr.8: 100522595-100522596 on Build GRCh38
rs779832755 Chr.8: 100522492-100522492 on Build GRCh38
rs767173630 Chr.8: 100522561-100522561 on Build GRCh38
rs1033489478 Chr.8: 100522527-100522527 on Build GRCh38
rs113970415 Chr.8: 100522536-100522536 on Build GRCh38
rs764351431 Chr.8: 100522554-100522554 on Build GRCh38
rs16898488 Chr.8: 100522505-100522505 on Build GRCh38
rs759003595 Chr.8: 100522636-100522636 on Build GRCh38
rs753338330 Chr.8: 100522627-100522627 on Build GRCh38
rs199719979 Chr.8: 100522584-100522584 on Build GRCh38
rs192246523 Chr.8: 100522519-100522519 on Build GRCh38
rs780640944 Chr.8: 100522641-100522641 on Build GRCh38
rs575770928 Chr.8: 100522600-100522600 on Build GRCh38
rs898274679 Chr.8: 100522475-100522475 on Build GRCh38
rs747652596 Chr.8: 100522596-100522596 on Build GRCh38
rs769827601 Chr.8: 100522531-100522531 on Build GRCh38
rs958363709 Chr.8: 100522569-100522569 on Build GRCh38
rs1041304686 Chr.8: 100522532-100522532 on Build GRCh38
rs774165489 Chr.8: 100522547-100522547 on Build GRCh38
rs372015313 Chr.8: 100522550-100522550 on Build GRCh38
rs773309514 Chr.8: 100522534-100522534 on Build GRCh38
rs182854174 Chr.8: 100522546-100522546 on Build GRCh38

View Full Product Details