Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105176749-105177022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
UniGene
Hs.506603
Gene Chromosome Location
Chr.12: 105173297-105236230 on Build GRCh38
Transcripts
NM_001251904.1 NM_001251905.1 NM_018171.3 XM_006719472.1 XM_011538530.2 XM_011538531.2 XM_011538532.2 XM_017019551.1 XM_017019552.1 XM_017019553.1 XM_017019554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5422135 c.1672-6C>A p.? Chr.12: 105177022-105177022 on Build GRCh38
2064875 c.1765G>A p.E589K Chr.12: 105176923-105176923 on Build GRCh38
934304 c.1741C>T p.R581C Chr.12: 105176947-105176947 on Build GRCh38
1183286 c.1682C>A p.T561N Chr.12: 105177006-105177006 on Build GRCh38
934303 c.1781A>G p.Y594C Chr.12: 105176907-105176907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761544638 Chr.12: 105176771-105176771 on Build GRCh38
rs538776087 Chr.12: 105176792-105176792 on Build GRCh38
rs776979017 Chr.12: 105176916-105176917 on Build GRCh38
rs555725549 Chr.12: 105176785-105176785 on Build GRCh38
rs765293784 Chr.12: 105176923-105176923 on Build GRCh38
rs752593594 Chr.12: 105176944-105176944 on Build GRCh38
rs373485718 Chr.12: 105176846-105176846 on Build GRCh38
rs949743544 Chr.12: 105176889-105176889 on Build GRCh38
rs758730830 Chr.12: 105176973-105176973 on Build GRCh38
rs767297070 Chr.12: 105176929-105176929 on Build GRCh38
rs781552911 Chr.12: 105176996-105176996 on Build GRCh38
rs199559987 Chr.12: 105176891-105176891 on Build GRCh38
rs370254990 Chr.12: 105176963-105176963 on Build GRCh38
rs779187749 Chr.12: 105176947-105176947 on Build GRCh38
rs759283532 Chr.12: 105176913-105176913 on Build GRCh38
rs756017295 Chr.12: 105176946-105176946 on Build GRCh38
rs747203652 Chr.12: 105176992-105176992 on Build GRCh38
rs761995028 Chr.12: 105176915-105176916 on Build GRCh38
rs780452705 Chr.12: 105176984-105176984 on Build GRCh38
rs750468282 Chr.12: 105176979-105176981 on Build GRCh38
rs184843985 Chr.12: 105176821-105176821 on Build GRCh38
rs1047495707 Chr.12: 105176870-105176870 on Build GRCh38
rs780610422 Chr.12: 105176843-105176843 on Build GRCh38
rs756070695 Chr.12: 105176905-105176905 on Build GRCh38
rs940772285 Chr.12: 105176951-105176951 on Build GRCh38
rs769298089 Chr.12: 105176887-105176887 on Build GRCh38
rs777023806 Chr.12: 105176881-105176881 on Build GRCh38
rs769011584 Chr.12: 105176904-105176907 on Build GRCh38
rs775875847 Chr.12: 105176970-105176971 on Build GRCh38
rs780899503 Chr.12: 105176819-105176819 on Build GRCh38
rs768914856 Chr.12: 105176994-105176994 on Build GRCh38
rs963681143 Chr.12: 105176833-105176833 on Build GRCh38
rs920604115 Chr.12: 105176983-105176983 on Build GRCh38
rs377356032 Chr.12: 105176825-105176825 on Build GRCh38
rs765995069 Chr.12: 105176899-105176899 on Build GRCh38
rs767273812 Chr.12: 105176801-105176801 on Build GRCh38
rs762652229 Chr.12: 105176896-105176896 on Build GRCh38
rs769022744 Chr.12: 105176871-105176871 on Build GRCh38

View Full Product Details