Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3753774-3754043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA3
Entrez Gene ID
9546
Gene Name
amyloid beta precursor protein binding family A member 3
Gene Aliases
MGC:15815, X11L2, mint3
UniGene
Hs.25527
Gene Chromosome Location
Chr.19: 3750773-3761697 on Build GRCh38
Transcripts
NM_004886.3 XM_006722950.3 XM_006722951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601882 c.970C>T p.R324C Chr.19: 3753806-3753806 on Build GRCh38
3362822 c.846C>G p.S282S Chr.19: 3754022-3754022 on Build GRCh38
6601881 c.971G>A p.R324H Chr.19: 3753805-3753805 on Build GRCh38
7278482 c.926G>A p.R309Q Chr.19: 3753850-3753850 on Build GRCh38
5644829 c.867C>T p.H289H Chr.19: 3753909-3753909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993661027 Chr.19: 3753974-3753974 on Build GRCh38
rs202130821 Chr.19: 3753809-3753809 on Build GRCh38
rs747171745 Chr.19: 3753923-3753923 on Build GRCh38
rs983581481 Chr.19: 3754020-3754020 on Build GRCh38
rs570833227 Chr.19: 3753850-3753850 on Build GRCh38
rs945836325 Chr.19: 3753911-3753911 on Build GRCh38
rs756881735 Chr.19: 3754014-3754014 on Build GRCh38
rs962160196 Chr.19: 3753968-3753968 on Build GRCh38
rs767228548 Chr.19: 3754006-3754006 on Build GRCh38
rs367758435 Chr.19: 3754001-3754001 on Build GRCh38
rs754517986 Chr.19: 3753847-3753847 on Build GRCh38
rs748717776 Chr.19: 3753998-3753998 on Build GRCh38
rs753830017 Chr.19: 3753963-3753963 on Build GRCh38
rs773406615 Chr.19: 3753903-3753903 on Build GRCh38
rs776507332 Chr.19: 3753996-3753996 on Build GRCh38
rs373142621 Chr.19: 3753853-3753853 on Build GRCh38
rs761296824 Chr.19: 3753826-3753826 on Build GRCh38
rs201224433 Chr.19: 3753933-3753933 on Build GRCh38
rs780418293 Chr.19: 3753984-3753984 on Build GRCh38
rs377689559 Chr.19: 3753898-3753898 on Build GRCh38
rs770149370 Chr.19: 3753999-3753999 on Build GRCh38
rs905758361 Chr.19: 3753805-3753805 on Build GRCh38
rs759204503 Chr.19: 3753908-3753908 on Build GRCh38
rs34868972 Chr.19: 3753876-3753876 on Build GRCh38
rs747143405 Chr.19: 3753988-3753988 on Build GRCh38
rs918111775 Chr.19: 3753835-3753835 on Build GRCh38
rs376259600 Chr.19: 3753844-3753844 on Build GRCh38
rs746107677 Chr.19: 3753869-3753869 on Build GRCh38
rs754193360 Chr.19: 3753836-3753836 on Build GRCh38
rs1016398997 Chr.19: 3753965-3753965 on Build GRCh38
rs377252138 Chr.19: 3753975-3753975 on Build GRCh38
rs370070756 Chr.19: 3754018-3754018 on Build GRCh38
rs200303476 Chr.19: 3753818-3753818 on Build GRCh38
rs901580965 Chr.19: 3753931-3753931 on Build GRCh38
rs937204747 Chr.19: 3753882-3753882 on Build GRCh38
rs1020254320 Chr.19: 3753913-3753913 on Build GRCh38
rs769675322 Chr.19: 3753806-3753806 on Build GRCh38
rs371868349 Chr.19: 3753829-3753829 on Build GRCh38
rs980781903 Chr.19: 3753865-3753865 on Build GRCh38
rs772292814 Chr.19: 3753821-3753821 on Build GRCh38
rs769353061 Chr.19: 3753881-3753881 on Build GRCh38
rs374545158 Chr.19: 3753894-3753894 on Build GRCh38
rs771081780 Chr.19: 3753939-3753940 on Build GRCh38
rs369277358 Chr.19: 3753852-3753852 on Build GRCh38
rs764079380 Chr.19: 3753934-3753934 on Build GRCh38
rs751236971 Chr.19: 3753845-3753845 on Build GRCh38
rs778518140 Chr.19: 3753867-3753867 on Build GRCh38
rs370443318 Chr.19: 3753976-3753976 on Build GRCh38
rs772386240 Chr.19: 3753870-3753870 on Build GRCh38
rs774631510 Chr.19: 3753813-3753813 on Build GRCh38
rs374251473 Chr.19: 3753964-3753964 on Build GRCh38
rs777660238 Chr.19: 3753851-3753851 on Build GRCh38
rs868772901 Chr.19: 3753884-3753884 on Build GRCh38
rs778801082 Chr.19: 3753969-3753969 on Build GRCh38
rs1020202330 Chr.19: 3753799-3753799 on Build GRCh38
rs763269469 Chr.19: 3754002-3754002 on Build GRCh38
rs768868526 Chr.19: 3753989-3753989 on Build GRCh38
rs775115362 Chr.19: 3754015-3754015 on Build GRCh38
rs561702584 Chr.19: 3753812-3753812 on Build GRCh38
rs1041882305 Chr.19: 3753920-3753920 on Build GRCh38
rs61731066 Chr.19: 3754022-3754022 on Build GRCh38
rs780709838 Chr.19: 3753848-3753848 on Build GRCh38
rs370997550 Chr.19: 3753909-3753909 on Build GRCh38
rs1050093519 Chr.19: 3753957-3753957 on Build GRCh38
rs760267251 Chr.19: 3754016-3754016 on Build GRCh38
rs752247384 Chr.19: 3753914-3753914 on Build GRCh38
rs372188071 Chr.19: 3753808-3753808 on Build GRCh38
rs867803530 Chr.19: 3753817-3753817 on Build GRCh38
rs775646823 Chr.19: 3753824-3753824 on Build GRCh38
rs906536055 Chr.19: 3753825-3753825 on Build GRCh38
rs374472856 Chr.19: 3753982-3753982 on Build GRCh38

View Full Product Details