Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2251105-2251354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMH
Entrez Gene ID
268
Gene Name
anti-Mullerian hormone
Gene Aliases
MIF, MIS
UniGene
Hs.112432
Gene Chromosome Location
Chr.19: 2249114-2252073 on Build GRCh38
Transcripts
NM_000479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195576 c.1027G>C p.E343Q Chr.19: 2251301-2251301 on Build GRCh38
5352206 c.1035_1037delGCT p.L350delL Chr.19: 2251309-2251311 on Build GRCh38
5434681 c.841G>C p.E281Q Chr.19: 2251115-2251115 on Build GRCh38
4131578 c.974A>G p.Q325R Chr.19: 2251248-2251248 on Build GRCh38
5747959 c.884C>T p.T295M Chr.19: 2251158-2251158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769350289 Chr.19: 2251124-2251124 on Build GRCh38
rs908446977 Chr.19: 2251130-2251130 on Build GRCh38
rs922145059 Chr.19: 2251223-2251223 on Build GRCh38
rs932711550 Chr.19: 2251164-2251164 on Build GRCh38
rs752466718 Chr.19: 2251183-2251183 on Build GRCh38
rs751238247 Chr.19: 2251284-2251285 on Build GRCh38
rs773772660 Chr.19: 2251268-2251268 on Build GRCh38
rs1032576781 Chr.19: 2251172-2251172 on Build GRCh38
rs750923515 Chr.19: 2251204-2251204 on Build GRCh38
rs1044973290 Chr.19: 2251299-2251299 on Build GRCh38
rs577002391 Chr.19: 2251241-2251241 on Build GRCh38
rs548613810 Chr.19: 2251209-2251209 on Build GRCh38
rs1014988797 Chr.19: 2251324-2251324 on Build GRCh38
rs953002502 Chr.19: 2251238-2251238 on Build GRCh38
rs1023152184 Chr.19: 2251258-2251258 on Build GRCh38
rs566806768 Chr.19: 2251224-2251224 on Build GRCh38
rs1041428114 Chr.19: 2251254-2251254 on Build GRCh38
rs779193966 Chr.19: 2251193-2251193 on Build GRCh38
rs893605605 Chr.19: 2251289-2251289 on Build GRCh38
rs758405250 Chr.19: 2251180-2251180 on Build GRCh38
rs775298247 Chr.19: 2251288-2251288 on Build GRCh38
rs556078854 Chr.19: 2251265-2251265 on Build GRCh38
rs962080364 Chr.19: 2251215-2251215 on Build GRCh38
rs994991215 Chr.19: 2251333-2251333 on Build GRCh38
rs199831511 Chr.19: 2251138-2251138 on Build GRCh38
rs759439167 Chr.19: 2251169-2251169 on Build GRCh38
rs757014694 Chr.19: 2251309-2251311 on Build GRCh38
rs774257374 Chr.19: 2251150-2251150 on Build GRCh38
rs536688211 Chr.19: 2251179-2251179 on Build GRCh38
rs764425903 Chr.19: 2251184-2251184 on Build GRCh38
rs981227702 Chr.19: 2251272-2251272 on Build GRCh38
rs908417518 Chr.19: 2251275-2251275 on Build GRCh38
rs757410331 Chr.19: 2251188-2251188 on Build GRCh38
rs764049634 Chr.19: 2251328-2251328 on Build GRCh38
rs1032678120 Chr.19: 2251196-2251196 on Build GRCh38
rs765937994 Chr.19: 2251144-2251144 on Build GRCh38
rs1003233694 Chr.19: 2251309-2251309 on Build GRCh38
rs747809683 Chr.19: 2251123-2251123 on Build GRCh38
rs140765565 Chr.19: 2251248-2251248 on Build GRCh38
rs1012178126 Chr.19: 2251290-2251290 on Build GRCh38
rs906216580 Chr.19: 2251308-2251309 on Build GRCh38
rs1021157391 Chr.19: 2251185-2251185 on Build GRCh38
rs772647922 Chr.19: 2251134-2251134 on Build GRCh38
rs770404493 Chr.19: 2251266-2251266 on Build GRCh38
rs759477280 Chr.19: 2251269-2251269 on Build GRCh38
rs558432762 Chr.19: 2251239-2251239 on Build GRCh38
rs1041109427 Chr.19: 2251127-2251127 on Build GRCh38
rs771925752 Chr.19: 2251287-2251287 on Build GRCh38
rs911237166 Chr.19: 2251245-2251245 on Build GRCh38
rs534325383 Chr.19: 2251230-2251230 on Build GRCh38
rs760391803 Chr.19: 2251298-2251298 on Build GRCh38
rs898529503 Chr.19: 2251153-2251153 on Build GRCh38
rs372200369 Chr.19: 2251199-2251199 on Build GRCh38
rs753904327 Chr.19: 2251187-2251187 on Build GRCh38

View Full Product Details