Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115841951-115842190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S1
Entrez Gene ID
1176
Gene Name
adaptor related protein complex 3 sigma 1 subunit
Gene Aliases
CLAPS3, Sigma3A
UniGene
Hs.406191
Gene Chromosome Location
Chr.5: 115841606-115914081 on Build GRCh38
Transcripts
NM_001002924.2 NM_001284.3 NM_001318090.1 NM_001318091.1 NM_001318093.1 NM_001318094.1 XM_011543136.2 XM_017009024.1 XM_017009025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323065 c.42G>A p.P14P Chr.5: 115842079-115842079 on Build GRCh38
299059 c.30C>A p.N10K Chr.5: 115842067-115842067 on Build GRCh38
6043875 c.64C>A p.P22T Chr.5: 115842101-115842101 on Build GRCh38
4422834 c.69_69+1insA p.S24fs*2 Chr.5: 115842106-115842107 on Build GRCh38
393689 c.39G>T p.K13N Chr.5: 115842076-115842076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768202808 Chr.5: 115842114-115842114 on Build GRCh38
rs771888426 Chr.5: 115841995-115841995 on Build GRCh38
rs779757861 Chr.5: 115842044-115842044 on Build GRCh38
rs764030106 Chr.5: 115842026-115842026 on Build GRCh38
rs200347946 Chr.5: 115842071-115842071 on Build GRCh38
rs773033432 Chr.5: 115842004-115842004 on Build GRCh38
rs11555658 Chr.5: 115842008-115842008 on Build GRCh38
rs572687640 Chr.5: 115841971-115841971 on Build GRCh38
rs369238013 Chr.5: 115842036-115842036 on Build GRCh38
rs775570888 Chr.5: 115842023-115842023 on Build GRCh38
rs377617825 Chr.5: 115842106-115842107 on Build GRCh38
rs771503181 Chr.5: 115842141-115842141 on Build GRCh38
rs748837438 Chr.5: 115842048-115842048 on Build GRCh38
rs754628724 Chr.5: 115842148-115842148 on Build GRCh38
rs141651630 Chr.5: 115842102-115842102 on Build GRCh38
rs368475651 Chr.5: 115842124-115842124 on Build GRCh38
rs11555657 Chr.5: 115841994-115841994 on Build GRCh38
rs752136463 Chr.5: 115842028-115842028 on Build GRCh38
rs113920317 Chr.5: 115842156-115842156 on Build GRCh38
rs539900902 Chr.5: 115841980-115841980 on Build GRCh38
rs977255252 Chr.5: 115842086-115842086 on Build GRCh38
rs374408873 Chr.5: 115842096-115842096 on Build GRCh38
rs927923307 Chr.5: 115841989-115841989 on Build GRCh38
rs756538558 Chr.5: 115842042-115842042 on Build GRCh38
rs746872767 Chr.5: 115842077-115842077 on Build GRCh38
rs935223106 Chr.5: 115841996-115841996 on Build GRCh38
rs1050864 Chr.5: 115842024-115842024 on Build GRCh38
rs770647592 Chr.5: 115842084-115842084 on Build GRCh38
rs370625498 Chr.5: 115842029-115842029 on Build GRCh38
rs750873511 Chr.5: 115842039-115842039 on Build GRCh38
rs760442918 Chr.5: 115842009-115842009 on Build GRCh38
rs754543487 Chr.5: 115842049-115842049 on Build GRCh38
rs548944937 Chr.5: 115842135-115842135 on Build GRCh38
rs1041580783 Chr.5: 115842149-115842149 on Build GRCh38
rs144226661 Chr.5: 115841971-115841972 on Build GRCh38
rs78864570 Chr.5: 115841975-115841976 on Build GRCh38
rs969279634 Chr.5: 115841982-115841982 on Build GRCh38
rs778608820 Chr.5: 115842155-115842155 on Build GRCh38
rs916642995 Chr.5: 115841981-115841981 on Build GRCh38
rs778248571 Chr.5: 115842064-115842064 on Build GRCh38
rs761967631 Chr.5: 115842111-115842111 on Build GRCh38
rs189493290 Chr.5: 115841990-115841990 on Build GRCh38
rs774362428 Chr.5: 115841988-115841988 on Build GRCh38
rs774342712 Chr.5: 115842107-115842107 on Build GRCh38
rs773771496 Chr.5: 115841992-115841992 on Build GRCh38
rs765503917 Chr.5: 115842019-115842019 on Build GRCh38
rs374131819 Chr.5: 115842068-115842068 on Build GRCh38
rs753562290 Chr.5: 115842143-115842143 on Build GRCh38
rs780601857 Chr.5: 115841975-115841975 on Build GRCh38
rs370092678 Chr.5: 115842085-115842085 on Build GRCh38
rs761131366 Chr.5: 115842132-115842132 on Build GRCh38
rs766607848 Chr.5: 115842139-115842139 on Build GRCh38
rs768890009 Chr.5: 115842103-115842103 on Build GRCh38
rs935726762 Chr.5: 115842126-115842126 on Build GRCh38
rs920921762 Chr.5: 115842160-115842160 on Build GRCh38

View Full Product Details