Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151118474-151118731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
UniGene
Hs.647075
Gene Chromosome Location
Chr.7: 151085831-151144436 on Build GRCh38
Transcripts
NM_001042535.3 NM_001281300.1 NM_001308304.1 NM_001308305.1 NM_031946.6 XM_005249943.2 XM_011515780.1 XM_017011732.1 XM_017011733.1 XM_017011734.1 XM_017011735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390318 c.849G>A p.Q283Q Chr.7: 151118512-151118512 on Build GRCh38
1182192 c.931G>A p.V311M Chr.7: 151118594-151118594 on Build GRCh38
6471760 c.874C>T p.Q292* Chr.7: 151118537-151118537 on Build GRCh38
7156536 c.911_922del12 p.P306_S309delPSHS Chr.7: 151118574-151118585 on Build GRCh38
1182197 c.914C>T p.S305L Chr.7: 151118577-151118577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755607014 Chr.7: 151118640-151118640 on Build GRCh38
rs200887887 Chr.7: 151118609-151118609 on Build GRCh38
rs61741392 Chr.7: 151118618-151118618 on Build GRCh38
rs1005373176 Chr.7: 151118692-151118692 on Build GRCh38
rs776832196 Chr.7: 151118602-151118602 on Build GRCh38
rs370482661 Chr.7: 151118650-151118650 on Build GRCh38
rs765507429 Chr.7: 151118545-151118545 on Build GRCh38
rs780819924 Chr.7: 151118590-151118590 on Build GRCh38
rs200599566 Chr.7: 151118636-151118636 on Build GRCh38
rs371001392 Chr.7: 151118603-151118603 on Build GRCh38
rs768545330 Chr.7: 151118652-151118652 on Build GRCh38
rs963809078 Chr.7: 151118695-151118695 on Build GRCh38
rs149050258 Chr.7: 151118710-151118710 on Build GRCh38
rs773551103 Chr.7: 151118598-151118598 on Build GRCh38
rs372837701 Chr.7: 151118599-151118599 on Build GRCh38
rs201733562 Chr.7: 151118614-151118614 on Build GRCh38
rs776724709 Chr.7: 151118673-151118673 on Build GRCh38
rs62489621 Chr.7: 151118581-151118581 on Build GRCh38
rs747187463 Chr.7: 151118669-151118669 on Build GRCh38
rs895906452 Chr.7: 151118655-151118655 on Build GRCh38
rs770913985 Chr.7: 151118600-151118600 on Build GRCh38
rs61740089 Chr.7: 151118543-151118543 on Build GRCh38
rs116615381 Chr.7: 151118693-151118693 on Build GRCh38
rs778921766 Chr.7: 151118653-151118653 on Build GRCh38
rs760791916 Chr.7: 151118515-151118515 on Build GRCh38
rs547546154 Chr.7: 151118670-151118670 on Build GRCh38
rs777364527 Chr.7: 151118572-151118572 on Build GRCh38
rs373083934 Chr.7: 151118591-151118591 on Build GRCh38
rs41282739 Chr.7: 151118637-151118637 on Build GRCh38
rs763255239 Chr.7: 151118617-151118617 on Build GRCh38
rs773596061 Chr.7: 151118509-151118509 on Build GRCh38
rs186709855 Chr.7: 151118702-151118702 on Build GRCh38
rs746713071 Chr.7: 151118578-151118578 on Build GRCh38
rs768590477 Chr.7: 151118594-151118594 on Build GRCh38
rs754272615 Chr.7: 151118529-151118529 on Build GRCh38
rs756991712 Chr.7: 151118626-151118626 on Build GRCh38
rs753003462 Chr.7: 151118551-151118551 on Build GRCh38
rs759620660 Chr.7: 151118674-151118674 on Build GRCh38
rs975320034 Chr.7: 151118562-151118562 on Build GRCh38
rs747354868 Chr.7: 151118555-151118555 on Build GRCh38
rs780087136 Chr.7: 151118649-151118649 on Build GRCh38
rs766532891 Chr.7: 151118528-151118528 on Build GRCh38
rs564448780 Chr.7: 151118701-151118701 on Build GRCh38
rs769786956 Chr.7: 151118684-151118684 on Build GRCh38
rs569191956 Chr.7: 151118593-151118593 on Build GRCh38
rs778017253 Chr.7: 151118506-151118506 on Build GRCh38

View Full Product Details