Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108827120-108827381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNAD1
Entrez Gene ID
254268
Gene Name
AKNA domain containing 1
Gene Aliases
C1orf62
UniGene
Hs.729441
Gene Chromosome Location
Chr.1: 108815898-108858242 on Build GRCh38
Transcripts
NM_152763.4 NR_049760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231894 c.1934C>T p.P645L Chr.1: 108827207-108827207 on Build GRCh38
462535 c.1846A>T p.N616Y Chr.1: 108827295-108827295 on Build GRCh38
6305922 c.1845A>G p.Q615Q Chr.1: 108827296-108827296 on Build GRCh38
1127497 c.1849G>A p.V617M Chr.1: 108827292-108827292 on Build GRCh38
4907380 c.980A>G p.K327R Chr.1: 108827282-108827282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770834502 Chr.1: 108827201-108827201 on Build GRCh38
rs759324708 Chr.1: 108827219-108827219 on Build GRCh38
rs745843903 Chr.1: 108827312-108827312 on Build GRCh38
rs181784849 Chr.1: 108827320-108827320 on Build GRCh38
rs370949868 Chr.1: 108827319-108827319 on Build GRCh38
rs756204106 Chr.1: 108827267-108827267 on Build GRCh38
rs985647345 Chr.1: 108827151-108827151 on Build GRCh38
rs747826508 Chr.1: 108827178-108827178 on Build GRCh38
rs779178361 Chr.1: 108827177-108827177 on Build GRCh38
rs373032742 Chr.1: 108827307-108827307 on Build GRCh38
rs552190178 Chr.1: 108827150-108827150 on Build GRCh38
rs755015083 Chr.1: 108827266-108827266 on Build GRCh38
rs750595892 Chr.1: 108827261-108827261 on Build GRCh38
rs764087099 Chr.1: 108827217-108827218 on Build GRCh38
rs754374426 Chr.1: 108827276-108827276 on Build GRCh38
rs760231054 Chr.1: 108827192-108827192 on Build GRCh38
rs756617428 Chr.1: 108827163-108827163 on Build GRCh38
rs376391474 Chr.1: 108827191-108827191 on Build GRCh38
rs767815626 Chr.1: 108827255-108827255 on Build GRCh38
rs573002533 Chr.1: 108827346-108827346 on Build GRCh38
rs978682064 Chr.1: 108827143-108827143 on Build GRCh38
rs190125386 Chr.1: 108827317-108827317 on Build GRCh38
rs762177802 Chr.1: 108827237-108827237 on Build GRCh38
rs771661558 Chr.1: 108827181-108827181 on Build GRCh38
rs11102505 Chr.1: 108827169-108827169 on Build GRCh38
rs925876142 Chr.1: 108827305-108827305 on Build GRCh38
rs772929222 Chr.1: 108827168-108827169 on Build GRCh38
rs184775863 Chr.1: 108827227-108827227 on Build GRCh38
rs773577913 Chr.1: 108827339-108827339 on Build GRCh38
rs770479314 Chr.1: 108827310-108827310 on Build GRCh38
rs888382793 Chr.1: 108827298-108827298 on Build GRCh38
rs1027251764 Chr.1: 108827139-108827139 on Build GRCh38
rs766311134 Chr.1: 108827272-108827272 on Build GRCh38
rs145703408 Chr.1: 108827208-108827208 on Build GRCh38
rs375991430 Chr.1: 108827349-108827349 on Build GRCh38
rs11102506 Chr.1: 108827175-108827175 on Build GRCh38
rs757495699 Chr.1: 108827351-108827351 on Build GRCh38
rs777392868 Chr.1: 108827302-108827302 on Build GRCh38
rs779270486 Chr.1: 108827292-108827292 on Build GRCh38
rs758115565 Chr.1: 108827297-108827297 on Build GRCh38
rs775377275 Chr.1: 108827229-108827229 on Build GRCh38
rs7551421 Chr.1: 108827293-108827293 on Build GRCh38
rs143591479 Chr.1: 108827279-108827279 on Build GRCh38
rs879613038 Chr.1: 108827275-108827275 on Build GRCh38
rs753901143 Chr.1: 108827305-108827305 on Build GRCh38
rs760877918 Chr.1: 108827204-108827216 on Build GRCh38
rs775967546 Chr.1: 108827311-108827311 on Build GRCh38
rs769834975 Chr.1: 108827314-108827314 on Build GRCh38

View Full Product Details