Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114673908-114674145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD1
Entrez Gene ID
270
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
MAD, MADA, MMDD
UniGene
Hs.89570
Gene Chromosome Location
Chr.1: 114673098-114695618 on Build GRCh38
Transcripts
NM_000036.2 NM_001172626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32812 c.1898C>A p.P633H Chr.1: 114673985-114673985 on Build GRCh38
6327362 c.1958T>A p.L653Q Chr.1: 114674024-114674024 on Build GRCh38
7130397 c.1929G>A p.M643I Chr.1: 114673954-114673954 on Build GRCh38
5667293 c.1801-3C>T p.? Chr.1: 114674085-114674085 on Build GRCh38
4902599 c.1847C>T p.A616V Chr.1: 114674036-114674036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367934256 Chr.1: 114674033-114674033 on Build GRCh38
rs760814471 Chr.1: 114673959-114673959 on Build GRCh38
rs1039812205 Chr.1: 114674003-114674003 on Build GRCh38
rs771050676 Chr.1: 114674074-114674074 on Build GRCh38
rs569798404 Chr.1: 114674068-114674068 on Build GRCh38
rs886045094 Chr.1: 114673957-114673957 on Build GRCh38
rs182610890 Chr.1: 114673952-114673952 on Build GRCh38
rs767670102 Chr.1: 114674051-114674051 on Build GRCh38
rs942391395 Chr.1: 114674002-114674002 on Build GRCh38
rs34257411 Chr.1: 114674038-114674038 on Build GRCh38
rs776575127 Chr.1: 114673984-114673984 on Build GRCh38
rs772179953 Chr.1: 114673948-114673948 on Build GRCh38
rs141900642 Chr.1: 114674064-114674064 on Build GRCh38
rs763816383 Chr.1: 114674017-114674017 on Build GRCh38
rs138019598 Chr.1: 114674037-114674037 on Build GRCh38
rs587779374 Chr.1: 114673940-114673940 on Build GRCh38
rs565587413 Chr.1: 114674034-114674034 on Build GRCh38
rs765059820 Chr.1: 114673990-114673990 on Build GRCh38
rs778123642 Chr.1: 114674105-114674108 on Build GRCh38
rs547408933 Chr.1: 114673995-114673995 on Build GRCh38
rs748560344 Chr.1: 114674040-114674040 on Build GRCh38
rs759306857 Chr.1: 114673988-114673988 on Build GRCh38
rs371341711 Chr.1: 114674041-114674041 on Build GRCh38
rs376186860 Chr.1: 114673989-114673989 on Build GRCh38
rs919600835 Chr.1: 114674010-114674010 on Build GRCh38
rs752339283 Chr.1: 114673994-114673994 on Build GRCh38
rs775205237 Chr.1: 114674091-114674091 on Build GRCh38
rs148515775 Chr.1: 114674077-114674077 on Build GRCh38
rs748230667 Chr.1: 114674096-114674096 on Build GRCh38
rs756299631 Chr.1: 114674105-114674105 on Build GRCh38
rs748449616 Chr.1: 114673947-114673947 on Build GRCh38
rs776521726 Chr.1: 114674076-114674076 on Build GRCh38
rs762806741 Chr.1: 114674110-114674110 on Build GRCh38
rs34778674 Chr.1: 114673972-114673972 on Build GRCh38
rs750967670 Chr.1: 114674021-114674021 on Build GRCh38
rs762567932 Chr.1: 114673996-114673996 on Build GRCh38
rs371914821 Chr.1: 114673941-114673941 on Build GRCh38
rs34287100 Chr.1: 114673936-114673936 on Build GRCh38

View Full Product Details