Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109661600-109661853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs757488198 Chr.X: 109661667-109661667 on Build GRCh38
rs758373148 Chr.X: 109661714-109661714 on Build GRCh38
rs1022710958 Chr.X: 109661679-109661679 on Build GRCh38
rs376911017 Chr.X: 109661693-109661693 on Build GRCh38
rs904072959 Chr.X: 109661749-109661749 on Build GRCh38
rs778872096 Chr.X: 109661680-109661680 on Build GRCh38
rs369534226 Chr.X: 109661694-109661694 on Build GRCh38
rs892450408 Chr.X: 109661668-109661669 on Build GRCh38
rs776255176 Chr.X: 109661746-109661746 on Build GRCh38
rs1015447665 Chr.X: 109661769-109661769 on Build GRCh38
rs961336995 Chr.X: 109661785-109661785 on Build GRCh38
rs1012673011 Chr.X: 109661674-109661674 on Build GRCh38
rs745950054 Chr.X: 109661758-109661760 on Build GRCh38
rs952375676 Chr.X: 109661730-109661730 on Build GRCh38
rs122458138 Chr.X: 109661643-109661643 on Build GRCh38
rs797045216 Chr.X: 109661651-109661652 on Build GRCh38

View Full Product Details