Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 277200-277433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP1
Entrez Gene ID
52
Gene Name
acid phosphatase 1, soluble
Gene Aliases
HAAP, LMW-PTP
UniGene
Hs.558296
Gene Chromosome Location
Chr.2: 264869-278283 on Build GRCh38
Transcripts
NM_004300.3 NM_007099.3 NR_024080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936322 c.439G>A p.V147I Chr.2: 277266-277266 on Build GRCh38
6592424 c.443G>T p.R148M Chr.2: 277270-277270 on Build GRCh38
6091430 c.400-1G>T p.? Chr.2: 277226-277226 on Build GRCh38
1407306 c.447C>A p.C149* Chr.2: 277274-277274 on Build GRCh38
6188824 c.410C>G p.S137C Chr.2: 277237-277237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547572404 Chr.2: 277313-277313 on Build GRCh38
rs768165451 Chr.2: 277228-277228 on Build GRCh38
rs564638147 Chr.2: 277281-277281 on Build GRCh38
rs529906639 Chr.2: 277325-277325 on Build GRCh38
rs1008076100 Chr.2: 277398-277398 on Build GRCh38
rs759869670 Chr.2: 277354-277354 on Build GRCh38
rs1042064742 Chr.2: 277339-277339 on Build GRCh38
rs919964432 Chr.2: 277233-277233 on Build GRCh38
rs773656331 Chr.2: 277347-277347 on Build GRCh38
rs370285099 Chr.2: 277385-277385 on Build GRCh38
rs938882195 Chr.2: 277302-277302 on Build GRCh38
rs77927634 Chr.2: 277314-277314 on Build GRCh38
rs55709515 Chr.2: 277250-277250 on Build GRCh38
rs1000620127 Chr.2: 277372-277372 on Build GRCh38
rs903553360 Chr.2: 277371-277371 on Build GRCh38
rs148061534 Chr.2: 277283-277283 on Build GRCh38
rs746343841 Chr.2: 277237-277240 on Build GRCh38
rs751887778 Chr.2: 277255-277255 on Build GRCh38
rs980626999 Chr.2: 277270-277270 on Build GRCh38
rs201011167 Chr.2: 277286-277286 on Build GRCh38
rs746666231 Chr.2: 277322-277322 on Build GRCh38
rs754817583 Chr.2: 277298-277298 on Build GRCh38
rs201887597 Chr.2: 277350-277350 on Build GRCh38
rs768110291 Chr.2: 277323-277323 on Build GRCh38
rs765467335 Chr.2: 277247-277247 on Build GRCh38
rs35569198 Chr.2: 277237-277237 on Build GRCh38
rs185734863 Chr.2: 277265-277265 on Build GRCh38
rs34540815 Chr.2: 277229-277229 on Build GRCh38
rs774650894 Chr.2: 277352-277352 on Build GRCh38
rs767447123 Chr.2: 277355-277355 on Build GRCh38
rs375641056 Chr.2: 277249-277249 on Build GRCh38
rs889612434 Chr.2: 277390-277390 on Build GRCh38
rs528542266 Chr.2: 277282-277282 on Build GRCh38
rs770303461 Chr.2: 277344-277344 on Build GRCh38
rs372370647 Chr.2: 277277-277277 on Build GRCh38
rs755052951 Chr.2: 277263-277263 on Build GRCh38
rs116833857 Chr.2: 277326-277326 on Build GRCh38
rs192683684 Chr.2: 277252-277252 on Build GRCh38
rs150164591 Chr.2: 277292-277292 on Build GRCh38
rs762409559 Chr.2: 277241-277241 on Build GRCh38
rs763395204 Chr.2: 277348-277348 on Build GRCh38

View Full Product Details