Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10482022-10482279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD3
Entrez Gene ID
272
Gene Name
adenosine monophosphate deaminase 3
Gene Aliases
-
UniGene
Hs.501890
Gene Chromosome Location
Chr.11: 10450321-10507579 on Build GRCh38
Transcripts
NM_000480.2 NM_001025389.1 NM_001025390.1 NM_001172430.1 NM_001172431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981257 c.593C>T p.A198V Chr.11: 10482202-10482202 on Build GRCh38
2109242 c.500C>A p.A167D Chr.11: 10482109-10482109 on Build GRCh38
5371544 c.552G>A p.R184R Chr.11: 10482161-10482161 on Build GRCh38
2109244 c.549G>A p.P183P Chr.11: 10482158-10482158 on Build GRCh38
6599420 c.531C>T p.L177L Chr.11: 10482140-10482140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777496309 Chr.11: 10482233-10482233 on Build GRCh38
rs866275020 Chr.11: 10482176-10482176 on Build GRCh38
rs760772452 Chr.11: 10482135-10482135 on Build GRCh38
rs757637339 Chr.11: 10482109-10482109 on Build GRCh38
rs374707086 Chr.11: 10482050-10482050 on Build GRCh38
rs372011884 Chr.11: 10482161-10482161 on Build GRCh38
rs774729347 Chr.11: 10482053-10482053 on Build GRCh38
rs757178861 Chr.11: 10482236-10482236 on Build GRCh38
rs367749804 Chr.11: 10482157-10482157 on Build GRCh38
rs755709235 Chr.11: 10482159-10482159 on Build GRCh38
rs139917184 Chr.11: 10482128-10482128 on Build GRCh38
rs770866014 Chr.11: 10482130-10482130 on Build GRCh38
rs200180931 Chr.11: 10482108-10482108 on Build GRCh38
rs746644069 Chr.11: 10482113-10482113 on Build GRCh38
rs766237847 Chr.11: 10482063-10482063 on Build GRCh38
rs778716028 Chr.11: 10482048-10482048 on Build GRCh38
rs201639499 Chr.11: 10482120-10482120 on Build GRCh38
rs754355489 Chr.11: 10482140-10482140 on Build GRCh38
rs754636700 Chr.11: 10482151-10482151 on Build GRCh38
rs371419233 Chr.11: 10482044-10482044 on Build GRCh38
rs765752211 Chr.11: 10482197-10482197 on Build GRCh38
rs899958277 Chr.11: 10482250-10482250 on Build GRCh38
rs748797134 Chr.11: 10482234-10482234 on Build GRCh38
rs770060554 Chr.11: 10482257-10482257 on Build GRCh38
rs752415155 Chr.11: 10482208-10482208 on Build GRCh38
rs778935977 Chr.11: 10482242-10482242 on Build GRCh38
rs886047586 Chr.11: 10482121-10482121 on Build GRCh38
rs745741300 Chr.11: 10482251-10482251 on Build GRCh38
rs774075201 Chr.11: 10482187-10482187 on Build GRCh38
rs184537234 Chr.11: 10482184-10482184 on Build GRCh38
rs747673137 Chr.11: 10482115-10482115 on Build GRCh38
rs752821070 Chr.11: 10482095-10482095 on Build GRCh38
rs908852358 Chr.11: 10482146-10482146 on Build GRCh38
rs11042836 Chr.11: 10482189-10482189 on Build GRCh38
rs779278244 Chr.11: 10482110-10482110 on Build GRCh38
rs778219413 Chr.11: 10482099-10482099 on Build GRCh38
rs767502041 Chr.11: 10482086-10482086 on Build GRCh38
rs202051288 Chr.11: 10482193-10482193 on Build GRCh38
rs750786612 Chr.11: 10482150-10482150 on Build GRCh38
rs755280931 Chr.11: 10482248-10482249 on Build GRCh38
rs766605831 Chr.11: 10482203-10482203 on Build GRCh38
rs767753231 Chr.11: 10482133-10482133 on Build GRCh38
rs750793624 Chr.11: 10482201-10482201 on Build GRCh38
rs777511548 Chr.11: 10482195-10482195 on Build GRCh38
rs774133774 Chr.11: 10482131-10482131 on Build GRCh38
rs147454158 Chr.11: 10482075-10482075 on Build GRCh38
rs764108261 Chr.11: 10482136-10482136 on Build GRCh38
rs1041230084 Chr.11: 10482214-10482214 on Build GRCh38
rs779211791 Chr.11: 10482141-10482141 on Build GRCh38
rs576976365 Chr.11: 10482188-10482188 on Build GRCh38
rs780623304 Chr.11: 10482114-10482114 on Build GRCh38
rs747764407 Chr.11: 10482158-10482158 on Build GRCh38
rs16907852 Chr.11: 10482209-10482209 on Build GRCh38
rs762205591 Chr.11: 10482160-10482160 on Build GRCh38
rs756660564 Chr.11: 10482096-10482096 on Build GRCh38
rs776685491 Chr.11: 10482194-10482194 on Build GRCh38
rs147701905 Chr.11: 10482134-10482134 on Build GRCh38

View Full Product Details