Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218264953-218265226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAMP
Entrez Gene ID
14
Gene Name
angio associated migratory cell protein
Gene Aliases
-
UniGene
Hs.83347
Gene Chromosome Location
Chr.2: 218264129-218270209 on Build GRCh38
Transcripts
NM_001087.4 NM_001302545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999402 c.1171C>T p.R391C Chr.2: 218265078-218265078 on Build GRCh38
4133444 c.1165A>C p.T389P Chr.2: 218265084-218265084 on Build GRCh38
6267604 c.1103C>A p.A368E Chr.2: 218265146-218265146 on Build GRCh38
3798639 c.1183G>C p.D395H Chr.2: 218265066-218265066 on Build GRCh38
6192605 c.1172G>A p.R391H Chr.2: 218265077-218265077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968174625 Chr.2: 218265078-218265078 on Build GRCh38
rs141536817 Chr.2: 218264990-218264990 on Build GRCh38
rs774401016 Chr.2: 218265106-218265106 on Build GRCh38
rs1003484082 Chr.2: 218264988-218264988 on Build GRCh38
rs781190732 Chr.2: 218265202-218265202 on Build GRCh38
rs775381838 Chr.2: 218265001-218265001 on Build GRCh38
rs765949921 Chr.2: 218265157-218265157 on Build GRCh38
rs138734332 Chr.2: 218264980-218264980 on Build GRCh38
rs768398634 Chr.2: 218265016-218265016 on Build GRCh38
rs976373167 Chr.2: 218265146-218265146 on Build GRCh38
rs771938768 Chr.2: 218265113-218265113 on Build GRCh38
rs772981491 Chr.2: 218265042-218265042 on Build GRCh38
rs749244617 Chr.2: 218265009-218265009 on Build GRCh38
rs370889171 Chr.2: 218265166-218265166 on Build GRCh38
rs757475022 Chr.2: 218265185-218265185 on Build GRCh38
rs552622695 Chr.2: 218265059-218265059 on Build GRCh38
rs150748826 Chr.2: 218265087-218265087 on Build GRCh38
rs749982937 Chr.2: 218265075-218265087 on Build GRCh38
rs772715738 Chr.2: 218265136-218265136 on Build GRCh38
rs199681193 Chr.2: 218265203-218265203 on Build GRCh38
rs747048601 Chr.2: 218264986-218264986 on Build GRCh38
rs948836330 Chr.2: 218265052-218265052 on Build GRCh38
rs763198037 Chr.2: 218265175-218265175 on Build GRCh38
rs373809460 Chr.2: 218265101-218265101 on Build GRCh38
rs757277345 Chr.2: 218265091-218265091 on Build GRCh38
rs568215295 Chr.2: 218265107-218265107 on Build GRCh38
rs767060488 Chr.2: 218265040-218265040 on Build GRCh38
rs758856081 Chr.2: 218265086-218265086 on Build GRCh38
rs761467520 Chr.2: 218265029-218265029 on Build GRCh38
rs776289331 Chr.2: 218265159-218265159 on Build GRCh38
rs753016891 Chr.2: 218265077-218265077 on Build GRCh38
rs115942379 Chr.2: 218265182-218265182 on Build GRCh38
rs368048910 Chr.2: 218265105-218265105 on Build GRCh38
rs1056248286 Chr.2: 218265004-218265004 on Build GRCh38
rs749156612 Chr.2: 218265102-218265102 on Build GRCh38
rs765648962 Chr.2: 218265172-218265172 on Build GRCh38
rs867345695 Chr.2: 218265200-218265200 on Build GRCh38
rs774021722 Chr.2: 218265018-218265018 on Build GRCh38
rs765574346 Chr.2: 218265061-218265061 on Build GRCh38
rs748248112 Chr.2: 218265076-218265076 on Build GRCh38
rs146196986 Chr.2: 218265060-218265060 on Build GRCh38
rs531234075 Chr.2: 218265049-218265049 on Build GRCh38
rs751739644 Chr.2: 218265184-218265184 on Build GRCh38
rs546660239 Chr.2: 218265090-218265090 on Build GRCh38
rs139083943 Chr.2: 218265173-218265173 on Build GRCh38
rs756143831 Chr.2: 218265082-218265082 on Build GRCh38
rs760428054 Chr.2: 218265147-218265147 on Build GRCh38
rs939151798 Chr.2: 218264991-218264991 on Build GRCh38
rs781051240 Chr.2: 218264992-218264992 on Build GRCh38
rs370095659 Chr.2: 218265050-218265050 on Build GRCh38

View Full Product Details