Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20973721-20973970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412997 c.373G>T p.G125C Chr.22: 20973867-20973867 on Build GRCh38
7354953 c.290G>T p.G97V Chr.22: 20973784-20973784 on Build GRCh38
6367641 c.334T>A p.Y112N Chr.22: 20973846-20973846 on Build GRCh38
7354958 c.269G>A p.R90Q Chr.22: 20973763-20973763 on Build GRCh38
4699666 c.309C>T p.H103H Chr.22: 20973821-20973821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764715947 Chr.22: 20973851-20973851 on Build GRCh38
rs770691793 Chr.22: 20973832-20973832 on Build GRCh38
rs552413450 Chr.22: 20973821-20973821 on Build GRCh38
rs1006712600 Chr.22: 20973914-20973914 on Build GRCh38
rs932490046 Chr.22: 20973923-20973923 on Build GRCh38
rs372949879 Chr.22: 20973907-20973907 on Build GRCh38
rs757703610 Chr.22: 20973872-20973872 on Build GRCh38
rs757587817 Chr.22: 20973749-20973749 on Build GRCh38
rs776278563 Chr.22: 20973841-20973841 on Build GRCh38
rs752047647 Chr.22: 20973852-20973852 on Build GRCh38
rs1045497610 Chr.22: 20973752-20973752 on Build GRCh38
rs899782050 Chr.22: 20973909-20973909 on Build GRCh38
rs377274696 Chr.22: 20973762-20973762 on Build GRCh38
rs1051394043 Chr.22: 20973925-20973925 on Build GRCh38
rs756274075 Chr.22: 20973763-20973763 on Build GRCh38
rs998526162 Chr.22: 20973932-20973932 on Build GRCh38
rs576417164 Chr.22: 20973755-20973755 on Build GRCh38
rs890973575 Chr.22: 20973931-20973931 on Build GRCh38
rs762507118 Chr.22: 20973849-20973849 on Build GRCh38
rs1023210202 Chr.22: 20973839-20973839 on Build GRCh38
rs34371345 Chr.22: 20973787-20973788 on Build GRCh38
rs139566403 Chr.22: 20973889-20973889 on Build GRCh38
rs1016804547 Chr.22: 20973927-20973927 on Build GRCh38
rs764765760 Chr.22: 20973844-20973844 on Build GRCh38
rs367600638 Chr.22: 20973822-20973822 on Build GRCh38
rs148456772 Chr.22: 20973848-20973848 on Build GRCh38
rs73392563 Chr.22: 20973747-20973747 on Build GRCh38
rs777260937 Chr.22: 20973831-20973852 on Build GRCh38
rs149749799 Chr.22: 20973788-20973788 on Build GRCh38
rs531634411 Chr.22: 20973888-20973888 on Build GRCh38
rs375928890 Chr.22: 20973825-20973825 on Build GRCh38
rs371361440 Chr.22: 20973823-20973823 on Build GRCh38
rs750729885 Chr.22: 20973884-20973884 on Build GRCh38
rs767980238 Chr.22: 20973879-20973879 on Build GRCh38
rs151248970 Chr.22: 20973842-20973842 on Build GRCh38
rs962622649 Chr.22: 20973938-20973938 on Build GRCh38

View Full Product Details