Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161048605-161048844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898429 c.2343G>T p.E781D Chr.1: 161048678-161048678 on Build GRCh38
898430 c.2245G>T p.E749* Chr.1: 161048776-161048776 on Build GRCh38
898431 c.2182A>C p.S728R Chr.1: 161048839-161048839 on Build GRCh38
6603393 c.2178T>C p.A726A Chr.1: 161048843-161048843 on Build GRCh38
3477112 c.2231A>G p.D744G Chr.1: 161048790-161048790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373107765 Chr.1: 161048706-161048706 on Build GRCh38
rs778633183 Chr.1: 161048701-161048701 on Build GRCh38
rs953131517 Chr.1: 161048627-161048627 on Build GRCh38
rs926811102 Chr.1: 161048772-161048772 on Build GRCh38
rs752574619 Chr.1: 161048796-161048796 on Build GRCh38
rs373072388 Chr.1: 161048753-161048753 on Build GRCh38
rs369809062 Chr.1: 161048635-161048635 on Build GRCh38
rs200401268 Chr.1: 161048762-161048762 on Build GRCh38
rs754821279 Chr.1: 161048630-161048630 on Build GRCh38
rs922435922 Chr.1: 161048740-161048740 on Build GRCh38
rs769069190 Chr.1: 161048707-161048707 on Build GRCh38
rs975151998 Chr.1: 161048735-161048735 on Build GRCh38
rs777783938 Chr.1: 161048805-161048805 on Build GRCh38
rs376301120 Chr.1: 161048731-161048731 on Build GRCh38
rs748854826 Chr.1: 161048689-161048689 on Build GRCh38
rs952294388 Chr.1: 161048664-161048664 on Build GRCh38
rs748069988 Chr.1: 161048726-161048726 on Build GRCh38
rs751892317 Chr.1: 161048777-161048777 on Build GRCh38
rs745571102 Chr.1: 161048818-161048818 on Build GRCh38
rs867420651 Chr.1: 161048672-161048672 on Build GRCh38
rs201790759 Chr.1: 161048800-161048800 on Build GRCh38
rs753655862 Chr.1: 161048806-161048806 on Build GRCh38
rs770292771 Chr.1: 161048692-161048692 on Build GRCh38
rs759817018 Chr.1: 161048782-161048782 on Build GRCh38
rs780979775 Chr.1: 161048670-161048670 on Build GRCh38
rs61733353 Chr.1: 161048632-161048632 on Build GRCh38
rs781444880 Chr.1: 161048815-161048829 on Build GRCh38
rs763106466 Chr.1: 161048749-161048749 on Build GRCh38
rs972153305 Chr.1: 161048717-161048717 on Build GRCh38
rs778382590 Chr.1: 161048814-161048814 on Build GRCh38
rs139131986 Chr.1: 161048808-161048808 on Build GRCh38
rs751060130 Chr.1: 161048650-161048650 on Build GRCh38
rs756054003 Chr.1: 161048640-161048640 on Build GRCh38
rs201790027 Chr.1: 161048795-161048795 on Build GRCh38
rs751357522 Chr.1: 161048665-161048685 on Build GRCh38
rs377370667 Chr.1: 161048711-161048711 on Build GRCh38
rs755222428 Chr.1: 161048776-161048777 on Build GRCh38
rs777534325 Chr.1: 161048652-161048652 on Build GRCh38
rs949690285 Chr.1: 161048738-161048738 on Build GRCh38
rs766298821 Chr.1: 161048769-161048769 on Build GRCh38
rs762346577 Chr.1: 161048713-161048713 on Build GRCh38
rs983774533 Chr.1: 161048694-161048694 on Build GRCh38

View Full Product Details