Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87504272-87504519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB1
Entrez Gene ID
5243
Gene Name
ATP binding cassette subfamily B member 1
Gene Aliases
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1
UniGene
Hs.489033
Gene Chromosome Location
Chr.7: 87503863-87713323 on Build GRCh38
Transcripts
NM_000927.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673258 c.3649G>A p.A1217T Chr.7: 87504437-87504437 on Build GRCh38
197657 c.3751G>A p.V1251I Chr.7: 87504335-87504335 on Build GRCh38
5047576 c.3728T>G p.I1243R Chr.7: 87504358-87504358 on Build GRCh38
1673257 c.3800A>G p.Y1267C Chr.7: 87504286-87504286 on Build GRCh38
1452575 c.3674G>A p.R1225H Chr.7: 87504412-87504412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776490673 Chr.7: 87504434-87504434 on Build GRCh38
rs200645658 Chr.7: 87504396-87504396 on Build GRCh38
rs199868523 Chr.7: 87504393-87504393 on Build GRCh38
rs140995435 Chr.7: 87504440-87504440 on Build GRCh38
rs45456698 Chr.7: 87504335-87504335 on Build GRCh38
rs761559025 Chr.7: 87504336-87504336 on Build GRCh38
rs773942612 Chr.7: 87504481-87504481 on Build GRCh38
rs199912437 Chr.7: 87504404-87504404 on Build GRCh38
rs750505383 Chr.7: 87504423-87504423 on Build GRCh38
rs139750664 Chr.7: 87504377-87504377 on Build GRCh38
rs201578293 Chr.7: 87504388-87504388 on Build GRCh38
rs2235051 Chr.7: 87504339-87504339 on Build GRCh38
rs146398820 Chr.7: 87504369-87504369 on Build GRCh38
rs202003598 Chr.7: 87504453-87504453 on Build GRCh38
rs745902169 Chr.7: 87504423-87504423 on Build GRCh38
rs772971590 Chr.7: 87504355-87504355 on Build GRCh38
rs768562415 Chr.7: 87504331-87504331 on Build GRCh38
rs1044890354 Chr.7: 87504316-87504316 on Build GRCh38
rs201225658 Chr.7: 87504408-87504408 on Build GRCh38
rs114847673 Chr.7: 87504488-87504488 on Build GRCh38
rs200637194 Chr.7: 87504379-87504379 on Build GRCh38
rs957128606 Chr.7: 87504432-87504432 on Build GRCh38
rs759048576 Chr.7: 87504483-87504483 on Build GRCh38
rs563375387 Chr.7: 87504389-87504389 on Build GRCh38
rs142093374 Chr.7: 87504365-87504365 on Build GRCh38
rs762504646 Chr.7: 87504363-87504363 on Build GRCh38
rs757394498 Chr.7: 87504398-87504398 on Build GRCh38
rs771100707 Chr.7: 87504317-87504317 on Build GRCh38
rs35721439 Chr.7: 87504319-87504319 on Build GRCh38
rs150207051 Chr.7: 87504452-87504452 on Build GRCh38
rs28364274 Chr.7: 87504335-87504335 on Build GRCh38
rs772846826 Chr.7: 87504468-87504468 on Build GRCh38
rs779103120 Chr.7: 87504412-87504412 on Build GRCh38
rs41309231 Chr.7: 87504417-87504417 on Build GRCh38
rs1008462286 Chr.7: 87504402-87504402 on Build GRCh38
rs201462254 Chr.7: 87504399-87504399 on Build GRCh38
rs141856581 Chr.7: 87504411-87504411 on Build GRCh38
rs768613690 Chr.7: 87504425-87504425 on Build GRCh38
rs201609930 Chr.7: 87504340-87504340 on Build GRCh38
rs201089646 Chr.7: 87504368-87504368 on Build GRCh38
rs1013586488 Chr.7: 87504322-87504322 on Build GRCh38
rs767563447 Chr.7: 87504406-87504407 on Build GRCh38
rs755563103 Chr.7: 87504372-87504372 on Build GRCh38
rs149482536 Chr.7: 87504318-87504318 on Build GRCh38
rs765898153 Chr.7: 87504471-87504471 on Build GRCh38
rs368970424 Chr.7: 87504435-87504435 on Build GRCh38
rs143356931 Chr.7: 87504447-87504447 on Build GRCh38

View Full Product Details