Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62597845-62598106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL3
Entrez Gene ID
27329
Gene Name
angiopoietin like 3
Gene Aliases
ANG-5, ANGPT5, ANL3, FHBL2
UniGene
Hs.209153
Gene Chromosome Location
Chr.1: 62597487-62606305 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
UniGene
Hs.744927
Gene Chromosome Location
Chr.1: 62454726-62688368 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982916 c.365C>G p.S122* Chr.1: 62597931-62597931 on Build GRCh38
6339537 c.377G>T p.S126I Chr.1: 62597943-62597943 on Build GRCh38
1343801 c.278_280delAAG p.E96delE Chr.1: 62597844-62597846 on Build GRCh38
1602610 c.376A>G p.S126G Chr.1: 62597942-62597942 on Build GRCh38
6406499 c.429G>T p.E143D Chr.1: 62597995-62597995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777106117 Chr.1: 62597937-62597937 on Build GRCh38
rs772744707 Chr.1: 62597918-62597918 on Build GRCh38
rs770659249 Chr.1: 62598036-62598036 on Build GRCh38
rs771793802 Chr.1: 62597915-62597915 on Build GRCh38
rs746623729 Chr.1: 62598033-62598033 on Build GRCh38
rs72649574 Chr.1: 62598017-62598017 on Build GRCh38
rs776972189 Chr.1: 62598043-62598043 on Build GRCh38
rs375934830 Chr.1: 62597963-62597963 on Build GRCh38
rs765671478 Chr.1: 62598052-62598052 on Build GRCh38
rs769518625 Chr.1: 62597910-62597911 on Build GRCh38
rs749133407 Chr.1: 62597977-62597977 on Build GRCh38
rs540771692 Chr.1: 62597938-62597938 on Build GRCh38
rs398122988 Chr.1: 62597929-62597933 on Build GRCh38
rs768762188 Chr.1: 62597876-62597876 on Build GRCh38
rs749257607 Chr.1: 62597874-62597874 on Build GRCh38
rs1000693667 Chr.1: 62598035-62598035 on Build GRCh38
rs779009755 Chr.1: 62597990-62597990 on Build GRCh38
rs398122987 Chr.1: 62598005-62598008 on Build GRCh38
rs752758133 Chr.1: 62597950-62597950 on Build GRCh38
rs200785483 Chr.1: 62597951-62597951 on Build GRCh38
rs1042280900 Chr.1: 62597970-62597970 on Build GRCh38
rs759948701 Chr.1: 62598048-62598048 on Build GRCh38
rs775976787 Chr.1: 62598056-62598056 on Build GRCh38
rs773994616 Chr.1: 62597882-62597882 on Build GRCh38
rs747595139 Chr.1: 62597999-62597999 on Build GRCh38
rs372257803 Chr.1: 62598067-62598067 on Build GRCh38
rs764103387 Chr.1: 62598076-62598076 on Build GRCh38
rs766681247 Chr.1: 62597927-62597927 on Build GRCh38
rs755889096 Chr.1: 62597962-62597962 on Build GRCh38
rs751759402 Chr.1: 62597952-62597952 on Build GRCh38
rs550972659 Chr.1: 62598059-62598059 on Build GRCh38
rs761064945 Chr.1: 62597920-62597920 on Build GRCh38
rs754954896 Chr.1: 62597984-62597984 on Build GRCh38
rs761469232 Chr.1: 62597894-62597894 on Build GRCh38
rs751707642 Chr.1: 62598079-62598079 on Build GRCh38
rs771514856 Chr.1: 62598003-62598003 on Build GRCh38

View Full Product Details