Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134939217-134939488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
UniGene
Hs.486520
Gene Chromosome Location
Chr.6: 134917390-134950122 on Build GRCh38
Transcripts
NM_001193480.1 NM_022568.3 NM_170771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7261682 c.458C>A p.P153H Chr.6: 134939400-134939400 on Build GRCh38
1073587 c.541G>T p.E181* Chr.6: 134939317-134939317 on Build GRCh38
6250080 c.477C>A p.Y159* Chr.6: 134939381-134939381 on Build GRCh38
1073588 c.509C>T p.A170V Chr.6: 134939349-134939349 on Build GRCh38
6598322 c.591A>G p.A197A Chr.6: 134939267-134939267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138635600 Chr.6: 134939299-134939299 on Build GRCh38
rs878960840 Chr.6: 134939241-134939241 on Build GRCh38
rs776771674 Chr.6: 134939291-134939291 on Build GRCh38
rs868029756 Chr.6: 134939291-134939292 on Build GRCh38
rs976024321 Chr.6: 134939256-134939256 on Build GRCh38
rs201801976 Chr.6: 134939411-134939411 on Build GRCh38
rs76101175 Chr.6: 134939448-134939448 on Build GRCh38
rs772726575 Chr.6: 134939252-134939252 on Build GRCh38
rs983038510 Chr.6: 134939374-134939374 on Build GRCh38
rs779339434 Chr.6: 134939249-134939249 on Build GRCh38
rs779692245 Chr.6: 134939420-134939420 on Build GRCh38
rs769471734 Chr.6: 134939349-134939350 on Build GRCh38
rs750250670 Chr.6: 134939298-134939298 on Build GRCh38
rs755804167 Chr.6: 134939364-134939364 on Build GRCh38
rs760873672 Chr.6: 134939259-134939259 on Build GRCh38
rs747503084 Chr.6: 134939381-134939381 on Build GRCh38
rs966585013 Chr.6: 134939286-134939286 on Build GRCh38
rs773269640 Chr.6: 134939419-134939419 on Build GRCh38
rs776774469 Chr.6: 134939384-134939384 on Build GRCh38
rs780740276 Chr.6: 134939350-134939350 on Build GRCh38
rs200628448 Chr.6: 134939425-134939425 on Build GRCh38
rs141761190 Chr.6: 134939340-134939340 on Build GRCh38
rs776098698 Chr.6: 134939255-134939255 on Build GRCh38
rs755537501 Chr.6: 134939449-134939449 on Build GRCh38
rs763846763 Chr.6: 134939452-134939452 on Build GRCh38
rs920336778 Chr.6: 134939355-134939355 on Build GRCh38
rs758889098 Chr.6: 134939458-134939458 on Build GRCh38
rs780753175 Chr.6: 134939372-134939372 on Build GRCh38
rs762086550 Chr.6: 134939296-134939296 on Build GRCh38
rs368304856 Chr.6: 134939427-134939427 on Build GRCh38
rs766338919 Chr.6: 134939314-134939314 on Build GRCh38
rs1010241357 Chr.6: 134939265-134939265 on Build GRCh38
rs776247598 Chr.6: 134939434-134939434 on Build GRCh38
rs768831731 Chr.6: 134939288-134939288 on Build GRCh38
rs751469454 Chr.6: 134939334-134939334 on Build GRCh38
rs569177400 Chr.6: 134939297-134939297 on Build GRCh38
rs747814023 Chr.6: 134939244-134939244 on Build GRCh38
rs753516354 Chr.6: 134939456-134939456 on Build GRCh38
rs374755074 Chr.6: 134939250-134939250 on Build GRCh38
rs759466133 Chr.6: 134939429-134939429 on Build GRCh38
rs374622136 Chr.6: 134939428-134939428 on Build GRCh38
rs767804899 Chr.6: 134939402-134939402 on Build GRCh38
rs141805258 Chr.6: 134939399-134939399 on Build GRCh38
rs746264434 Chr.6: 134939367-134939367 on Build GRCh38
rs1020301593 Chr.6: 134939266-134939266 on Build GRCh38
rs758804155 Chr.6: 134939369-134939369 on Build GRCh38
rs144585433 Chr.6: 134939354-134939354 on Build GRCh38
rs368004380 Chr.6: 134939438-134939438 on Build GRCh38
rs771259375 Chr.6: 134939248-134939248 on Build GRCh38
rs777242234 Chr.6: 134939365-134939365 on Build GRCh38
rs768919719 Chr.6: 134939382-134939382 on Build GRCh38
rs148475686 Chr.6: 134939386-134939386 on Build GRCh38

View Full Product Details