Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6644283-6644546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592487 c.708G>T p.Q236H Chr.12: 6644373-6644373 on Build GRCh38
1476855 c.604G>A p.E202K Chr.12: 6644477-6644477 on Build GRCh38
6242766 c.587delA p.K196fs*80 Chr.12: 6644494-6644494 on Build GRCh38
3384574 c.561A>C p.G187G Chr.12: 6644520-6644520 on Build GRCh38
6592483 c.667G>T p.E223* Chr.12: 6644414-6644414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766563766 Chr.12: 6644420-6644420 on Build GRCh38
rs756181680 Chr.12: 6644432-6644433 on Build GRCh38
rs752696343 Chr.12: 6644518-6644518 on Build GRCh38
rs199906590 Chr.12: 6644357-6644357 on Build GRCh38
rs1021299682 Chr.12: 6644455-6644455 on Build GRCh38
rs761135961 Chr.12: 6644377-6644377 on Build GRCh38
rs567038756 Chr.12: 6644459-6644459 on Build GRCh38
rs758605145 Chr.12: 6644415-6644415 on Build GRCh38
rs773515146 Chr.12: 6644491-6644491 on Build GRCh38
rs752397473 Chr.12: 6644460-6644460 on Build GRCh38
rs201620539 Chr.12: 6644500-6644500 on Build GRCh38
rs781088084 Chr.12: 6644339-6644339 on Build GRCh38
rs552836560 Chr.12: 6644371-6644371 on Build GRCh38
rs142341550 Chr.12: 6644404-6644404 on Build GRCh38
rs868859399 Chr.12: 6644350-6644350 on Build GRCh38
rs751729156 Chr.12: 6644431-6644431 on Build GRCh38
rs777125457 Chr.12: 6644481-6644481 on Build GRCh38
rs770180945 Chr.12: 6644488-6644488 on Build GRCh38
rs535928418 Chr.12: 6644478-6644478 on Build GRCh38
rs773914526 Chr.12: 6644375-6644375 on Build GRCh38
rs146183012 Chr.12: 6644338-6644338 on Build GRCh38
rs745402029 Chr.12: 6644476-6644476 on Build GRCh38
rs772952796 Chr.12: 6644360-6644360 on Build GRCh38
rs368867411 Chr.12: 6644346-6644346 on Build GRCh38
rs748960906 Chr.12: 6644465-6644465 on Build GRCh38
rs376933199 Chr.12: 6644506-6644506 on Build GRCh38
rs750428930 Chr.12: 6644321-6644321 on Build GRCh38
rs532605296 Chr.12: 6644374-6644374 on Build GRCh38
rs765515031 Chr.12: 6644317-6644317 on Build GRCh38
rs777238087 Chr.12: 6644462-6644462 on Build GRCh38
rs779797390 Chr.12: 6644332-6644332 on Build GRCh38
rs750670447 Chr.12: 6644412-6644412 on Build GRCh38
rs778368526 Chr.12: 6644473-6644473 on Build GRCh38
rs771849510 Chr.12: 6644501-6644501 on Build GRCh38
rs139969601 Chr.12: 6644359-6644359 on Build GRCh38
rs764631874 Chr.12: 6644382-6644382 on Build GRCh38
rs763204631 Chr.12: 6644496-6644496 on Build GRCh38
rs776929000 Chr.12: 6644392-6644392 on Build GRCh38
rs770525894 Chr.12: 6644470-6644470 on Build GRCh38
rs558825862 Chr.12: 6644517-6644517 on Build GRCh38
rs373212885 Chr.12: 6644505-6644505 on Build GRCh38
rs762024158 Chr.12: 6644485-6644485 on Build GRCh38
rs748268820 Chr.12: 6644395-6644406 on Build GRCh38
rs758507910 Chr.12: 6644331-6644331 on Build GRCh38
rs137907685 Chr.12: 6644356-6644356 on Build GRCh38
rs146403423 Chr.12: 6644446-6644446 on Build GRCh38
rs1009457737 Chr.12: 6644367-6644367 on Build GRCh38
rs367723803 Chr.12: 6644461-6644461 on Build GRCh38

View Full Product Details