Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 118224698-118224933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD7
Entrez Gene ID
56311
Gene Name
ankyrin repeat domain 7
Gene Aliases
TSA806
UniGene
Hs.729340
Gene Chromosome Location
Chr.7: 118224658-118242731 on Build GRCh38
Transcripts
NM_019644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661953 c.67G>T p.E23* Chr.7: 118224897-118224897 on Build GRCh38
5811177 c.78A>C p.L26F Chr.7: 118224908-118224908 on Build GRCh38
1182962 c.5A>C p.N2T Chr.7: 118224835-118224835 on Build GRCh38
3877525 c.24G>T p.W8C Chr.7: 118224854-118224854 on Build GRCh38
4636500 c.65G>C p.R22P Chr.7: 118224895-118224895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201254477 Chr.7: 118224895-118224895 on Build GRCh38
rs764836515 Chr.7: 118224865-118224865 on Build GRCh38
rs758103577 Chr.7: 118224875-118224875 on Build GRCh38
rs770732142 Chr.7: 118224793-118224793 on Build GRCh38
rs748990895 Chr.7: 118224814-118224815 on Build GRCh38
rs1019745614 Chr.7: 118224775-118224775 on Build GRCh38
rs780083297 Chr.7: 118224907-118224907 on Build GRCh38
rs781048286 Chr.7: 118224890-118224890 on Build GRCh38
rs745877912 Chr.7: 118224802-118224802 on Build GRCh38
rs754710815 Chr.7: 118224820-118224836 on Build GRCh38
rs529203310 Chr.7: 118224725-118224725 on Build GRCh38
rs369714864 Chr.7: 118224827-118224827 on Build GRCh38
rs781336489 Chr.7: 118224796-118224796 on Build GRCh38
rs753405655 Chr.7: 118224831-118224831 on Build GRCh38
rs746987251 Chr.7: 118224786-118224786 on Build GRCh38
rs751243854 Chr.7: 118224879-118224879 on Build GRCh38
rs757231606 Chr.7: 118224889-118224889 on Build GRCh38
rs775751820 Chr.7: 118224809-118224809 on Build GRCh38
rs763127691 Chr.7: 118224814-118224814 on Build GRCh38
rs557885082 Chr.7: 118224900-118224900 on Build GRCh38
rs748507966 Chr.7: 118224837-118224837 on Build GRCh38
rs977480985 Chr.7: 118224844-118224844 on Build GRCh38
rs777653179 Chr.7: 118224877-118224877 on Build GRCh38
rs924601572 Chr.7: 118224854-118224854 on Build GRCh38
rs778517362 Chr.7: 118224841-118224841 on Build GRCh38
rs962308106 Chr.7: 118224743-118224743 on Build GRCh38
rs774533897 Chr.7: 118224825-118224825 on Build GRCh38
rs376379560 Chr.7: 118224871-118224871 on Build GRCh38
rs547668228 Chr.7: 118224808-118224808 on Build GRCh38
rs760199258 Chr.7: 118224902-118224902 on Build GRCh38
rs948556640 Chr.7: 118224903-118224903 on Build GRCh38
rs373642723 Chr.7: 118224828-118224828 on Build GRCh38
rs923423020 Chr.7: 118224774-118224774 on Build GRCh38
rs769054984 Chr.7: 118224817-118224817 on Build GRCh38
rs976250309 Chr.7: 118224746-118224746 on Build GRCh38
rs964148044 Chr.7: 118224789-118224789 on Build GRCh38
rs759175504 Chr.7: 118224845-118224845 on Build GRCh38
rs781650587 Chr.7: 118224741-118224741 on Build GRCh38
rs779741780 Chr.7: 118224805-118224806 on Build GRCh38

View Full Product Details