Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 60382375-60382602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA2
Entrez Gene ID
302
Gene Name
annexin A2
Gene Aliases
ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
UniGene
Hs.511605
Gene Chromosome Location
Chr.15: 60347151-60398025 on Build GRCh38
Transcripts
NM_001002857.1 NM_001002858.2 NM_001136015.2 NM_004039.2 XM_011521477.2 XM_017022090.1 XM_017022091.1 XM_017022092.1 XM_017022093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
385130 c.143A>G p.Y48C Chr.15: 60382401-60382401 on Build GRCh38
1194299 c.136A>C p.K46Q Chr.15: 60382408-60382408 on Build GRCh38
5849469 c.157G>A p.A53T Chr.15: 60382387-60382387 on Build GRCh38
112019 c.145_146insTATA p.T49fs*3 Chr.15: 60382398-60382399 on Build GRCh38
6569051 c.160G>A p.E54K Chr.15: 60382384-60382384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41304760 Chr.15: 60382457-60382457 on Build GRCh38
rs750751518 Chr.15: 60382474-60382474 on Build GRCh38
rs376828881 Chr.15: 60382456-60382456 on Build GRCh38
rs768653435 Chr.15: 60382409-60382409 on Build GRCh38
rs760092295 Chr.15: 60382495-60382495 on Build GRCh38
rs774177312 Chr.15: 60382449-60382449 on Build GRCh38
rs944876898 Chr.15: 60382423-60382423 on Build GRCh38
rs747064927 Chr.15: 60382401-60382401 on Build GRCh38
rs76472852 Chr.15: 60382524-60382524 on Build GRCh38
rs535738831 Chr.15: 60382498-60382498 on Build GRCh38
rs142107939 Chr.15: 60382548-60382548 on Build GRCh38
rs1043099006 Chr.15: 60382568-60382568 on Build GRCh38
rs41315968 Chr.15: 60382406-60382406 on Build GRCh38
rs747480107 Chr.15: 60382479-60382479 on Build GRCh38
rs376371711 Chr.15: 60382475-60382475 on Build GRCh38
rs767017960 Chr.15: 60382460-60382460 on Build GRCh38
rs919695794 Chr.15: 60382450-60382450 on Build GRCh38
rs752287988 Chr.15: 60382464-60382464 on Build GRCh38
rs3759911 Chr.15: 60382473-60382473 on Build GRCh38
rs574968041 Chr.15: 60382482-60382482 on Build GRCh38
rs142567732 Chr.15: 60382425-60382425 on Build GRCh38
rs904606185 Chr.15: 60382572-60382572 on Build GRCh38
rs368142886 Chr.15: 60382461-60382461 on Build GRCh38
rs781409432 Chr.15: 60382492-60382492 on Build GRCh38
rs1040991635 Chr.15: 60382444-60382444 on Build GRCh38
rs762696326 Chr.15: 60382430-60382430 on Build GRCh38
rs554090145 Chr.15: 60382513-60382513 on Build GRCh38
rs755748356 Chr.15: 60382471-60382471 on Build GRCh38
rs766318356 Chr.15: 60382432-60382432 on Build GRCh38
rs146367349 Chr.15: 60382550-60382550 on Build GRCh38
rs780461746 Chr.15: 60382398-60382398 on Build GRCh38
rs780574461 Chr.15: 60382477-60382477 on Build GRCh38
rs773138183 Chr.15: 60382429-60382429 on Build GRCh38
rs762143123 Chr.15: 60382422-60382422 on Build GRCh38
rs755031000 Chr.15: 60382487-60382487 on Build GRCh38
rs139842070 Chr.15: 60382556-60382556 on Build GRCh38
rs200817902 Chr.15: 60382397-60382397 on Build GRCh38
rs372107517 Chr.15: 60382478-60382478 on Build GRCh38

View Full Product Details