Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58183916-58184179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
UniGene
Hs.104624
Gene Chromosome Location
Chr.15: 58138169-58185911 on Build GRCh38
Transcripts
NM_001320635.1 NM_001320636.1 NM_020980.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4602443 c.757G>T p.V253F Chr.15: 58184004-58184004 on Build GRCh38
6210439 c.885G>A p.M295I Chr.15: 58184132-58184132 on Build GRCh38
1183305 c.861G>T p.E287D Chr.15: 58184108-58184108 on Build GRCh38
5362923 c.776G>T p.G259V Chr.15: 58184023-58184023 on Build GRCh38
7221315 c.835A>G p.T279A Chr.15: 58184082-58184082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1867380 Chr.15: 58184082-58184082 on Build GRCh38
rs749429402 Chr.15: 58184058-58184058 on Build GRCh38
rs779109780 Chr.15: 58184147-58184150 on Build GRCh38
rs749376493 Chr.15: 58183952-58183952 on Build GRCh38
rs779060680 Chr.15: 58184142-58184142 on Build GRCh38
rs774310850 Chr.15: 58183993-58183993 on Build GRCh38
rs761335431 Chr.15: 58184120-58184120 on Build GRCh38
rs753410663 Chr.15: 58184011-58184011 on Build GRCh38
rs759858030 Chr.15: 58183989-58183989 on Build GRCh38
rs778181894 Chr.15: 58184138-58184138 on Build GRCh38
rs182216973 Chr.15: 58184025-58184025 on Build GRCh38
rs762955710 Chr.15: 58184131-58184131 on Build GRCh38
rs374284559 Chr.15: 58184041-58184041 on Build GRCh38
rs745867789 Chr.15: 58184144-58184144 on Build GRCh38
rs764829595 Chr.15: 58184127-58184127 on Build GRCh38
rs761021408 Chr.15: 58184112-58184112 on Build GRCh38
rs74016588 Chr.15: 58184156-58184156 on Build GRCh38
rs1027229138 Chr.15: 58184069-58184069 on Build GRCh38
rs775782160 Chr.15: 58183998-58183998 on Build GRCh38
rs750410060 Chr.15: 58184032-58184032 on Build GRCh38
rs767955949 Chr.15: 58183996-58183996 on Build GRCh38
rs142822245 Chr.15: 58184044-58184044 on Build GRCh38
rs780523751 Chr.15: 58184147-58184147 on Build GRCh38
rs776240993 Chr.15: 58184118-58184118 on Build GRCh38
rs143394617 Chr.15: 58184005-58184005 on Build GRCh38
rs371099893 Chr.15: 58183990-58183990 on Build GRCh38
rs756473144 Chr.15: 58184045-58184045 on Build GRCh38
rs766626294 Chr.15: 58183997-58183997 on Build GRCh38
rs758224719 Chr.15: 58184033-58184033 on Build GRCh38
rs375847778 Chr.15: 58184017-58184017 on Build GRCh38
rs760535344 Chr.15: 58184141-58184141 on Build GRCh38
rs1002894992 Chr.15: 58184037-58184037 on Build GRCh38
rs749847313 Chr.15: 58184128-58184128 on Build GRCh38
rs778130721 Chr.15: 58184054-58184054 on Build GRCh38
rs140578393 Chr.15: 58184039-58184039 on Build GRCh38
rs771400420 Chr.15: 58183957-58183957 on Build GRCh38
rs763666523 Chr.15: 58184008-58184008 on Build GRCh38
rs778818501 Chr.15: 58184081-58184081 on Build GRCh38
rs769922791 Chr.15: 58184143-58184145 on Build GRCh38
rs774901177 Chr.15: 58183964-58183964 on Build GRCh38
rs766415923 Chr.15: 58184132-58184132 on Build GRCh38
rs374406557 Chr.15: 58184027-58184027 on Build GRCh38
rs201914936 Chr.15: 58184098-58184098 on Build GRCh38
rs367638674 Chr.15: 58184093-58184093 on Build GRCh38
rs377733309 Chr.15: 58184140-58184140 on Build GRCh38
rs751460577 Chr.15: 58184133-58184133 on Build GRCh38
rs771012823 Chr.15: 58184075-58184075 on Build GRCh38
rs772700269 Chr.15: 58184145-58184145 on Build GRCh38
rs868763994 Chr.15: 58184115-58184115 on Build GRCh38
rs768351351 Chr.15: 58184116-58184116 on Build GRCh38
rs748087825 Chr.15: 58184112-58184113 on Build GRCh38
rs756773366 Chr.15: 58184023-58184023 on Build GRCh38

View Full Product Details