Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138106543-138106788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1D1
Entrez Gene ID
6718
Gene Name
aldo-keto reductase family 1, member D1
Gene Aliases
3o5bred, CBAS2, SRD5B1
UniGene
Hs.201667
Gene Chromosome Location
Chr.7: 138076432-138118304 on Build GRCh38
Transcripts
NM_001190906.1 NM_001190907.1 NM_005989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309034 c.592C>G p.P198A Chr.7: 138106620-138106620 on Build GRCh38
1085986 c.623T>G p.F208C Chr.7: 138106651-138106651 on Build GRCh38
5871925 c.661C>T p.P221S Chr.7: 138106689-138106689 on Build GRCh38
3633834 c.600C>T p.F200F Chr.7: 138106628-138106628 on Build GRCh38
2859767 c.659G>A p.S220N Chr.7: 138106687-138106687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543945550 Chr.7: 138106739-138106739 on Build GRCh38
rs201408607 Chr.7: 138106683-138106683 on Build GRCh38
rs778994672 Chr.7: 138106662-138106662 on Build GRCh38
rs769255574 Chr.7: 138106576-138106576 on Build GRCh38
rs367883938 Chr.7: 138106722-138106722 on Build GRCh38
rs766498724 Chr.7: 138106628-138106628 on Build GRCh38
rs114392001 Chr.7: 138106624-138106624 on Build GRCh38
rs762292527 Chr.7: 138106738-138106738 on Build GRCh38
rs768395455 Chr.7: 138106728-138106728 on Build GRCh38
rs121918342 Chr.7: 138106621-138106621 on Build GRCh38
rs958292013 Chr.7: 138106572-138106572 on Build GRCh38
rs745742597 Chr.7: 138106706-138106706 on Build GRCh38
rs755282345 Chr.7: 138106643-138106643 on Build GRCh38
rs974928831 Chr.7: 138106588-138106588 on Build GRCh38
rs114135471 Chr.7: 138106597-138106597 on Build GRCh38
rs575025641 Chr.7: 138106718-138106718 on Build GRCh38
rs781611796 Chr.7: 138106672-138106672 on Build GRCh38
rs773858110 Chr.7: 138106749-138106749 on Build GRCh38
rs374974593 Chr.7: 138106663-138106663 on Build GRCh38
rs1025997766 Chr.7: 138106665-138106665 on Build GRCh38
rs372065794 Chr.7: 138106753-138106753 on Build GRCh38
rs762613714 Chr.7: 138106595-138106595 on Build GRCh38
rs754019534 Chr.7: 138106635-138106635 on Build GRCh38
rs749744865 Chr.7: 138106607-138106607 on Build GRCh38
rs781141323 Chr.7: 138106685-138106685 on Build GRCh38
rs766781831 Chr.7: 138106759-138106759 on Build GRCh38
rs769310676 Chr.7: 138106714-138106714 on Build GRCh38
rs754786571 Chr.7: 138106573-138106573 on Build GRCh38
rs759500212 Chr.7: 138106761-138106761 on Build GRCh38
rs768190295 Chr.7: 138106622-138106622 on Build GRCh38
rs371843537 Chr.7: 138106594-138106594 on Build GRCh38
rs754012632 Chr.7: 138106760-138106760 on Build GRCh38
rs940741321 Chr.7: 138106678-138106678 on Build GRCh38

View Full Product Details