The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014289.3 | NP_055104.2 | 2 - 3 | 359 | 69 |
GenBank mRNA | AF029232.1 | - | 2 - 3 | 259 | 69 |
AJ000388.1 | - | 2 - 3 | 350 | 69 | |
AK300340.1 | - | 1 - 2 | 108 | 69 | |
AK312701.1 | - | 2 - 3 | 323 | 69 | |
BC000730.2 | - | 1 - 2 | 188 | 69 | |
DA863483.1 | - | 2 - 3 | 359 | 69 |