The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014289.3 | NP_055104.2 | 6 - 7 | 1086 | 82 |
GenBank mRNA | AF029232.1 | - | 6 - 7 | 986 | 82 |
AJ000388.1 | - | 6 - 7 | 1077 | 82 | |
AK300340.1 | - | 4 - 5 | 642 | 82 | |
AK312701.1 | - | 6 - 7 | 1050 | 82 | |
BC000730.2 | - | 5 - 6 | 915 | 82 |