The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014289.3 | NP_055104.2 | 7 - 8 | 1161 | 67 |
GenBank mRNA | AF029232.1 | - | 7 - 8 | 1061 | 67 |
AJ000388.1 | - | 7 - 8 | 1152 | 67 | |
AK300340.1 | - | 5 - 6 | 717 | 67 | |
AK312701.1 | - | 7 - 8 | 1125 | 67 | |
BC000730.2 | - | 6 - 7 | 990 | 67 |