Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31077964-31078225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCYAP1R1
Entrez Gene ID
117
Gene Name
ADCYAP receptor type I
Gene Aliases
PAC1, PAC1R, PACAPR, PACAPRI
UniGene
Hs.377783
Gene Chromosome Location
Chr.7: 31052341-31114005 on Build GRCh38
Transcripts
NM_001118.4 NM_001199635.1 NM_001199636.1 NM_001199637.1 XM_005249618.4 XM_006715645.2 XM_017011736.1 XM_017011737.1 XM_017011738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5872593 c.211G>A p.E71K Chr.7: 31078044-31078044 on Build GRCh38
373269 c.166G>T p.G56W Chr.7: 31077999-31077999 on Build GRCh38
6269478 c.186G>T p.T62T Chr.7: 31078019-31078019 on Build GRCh38
5835398 c.263delA p.V89fs*18 Chr.7: 31078096-31078096 on Build GRCh38
3227560 c.164C>T p.P55L Chr.7: 31077997-31077997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746524853 Chr.7: 31078139-31078139 on Build GRCh38
rs374198118 Chr.7: 31078128-31078128 on Build GRCh38
rs188230760 Chr.7: 31078206-31078206 on Build GRCh38
rs746719200 Chr.7: 31078037-31078037 on Build GRCh38
rs140775607 Chr.7: 31078032-31078032 on Build GRCh38
rs146852802 Chr.7: 31078019-31078019 on Build GRCh38
rs140643573 Chr.7: 31078127-31078127 on Build GRCh38
rs752475879 Chr.7: 31078043-31078043 on Build GRCh38
rs769047161 Chr.7: 31078072-31078072 on Build GRCh38
rs769874446 Chr.7: 31078009-31078011 on Build GRCh38
rs749681357 Chr.7: 31078060-31078060 on Build GRCh38
rs753693167 Chr.7: 31078018-31078018 on Build GRCh38
rs935809690 Chr.7: 31078184-31078184 on Build GRCh38
rs994358999 Chr.7: 31078016-31078016 on Build GRCh38
rs759530805 Chr.7: 31078117-31078117 on Build GRCh38
rs1000835880 Chr.7: 31078033-31078033 on Build GRCh38
rs967834773 Chr.7: 31078159-31078159 on Build GRCh38
rs911734317 Chr.7: 31078175-31078175 on Build GRCh38
rs562824556 Chr.7: 31078054-31078054 on Build GRCh38
rs202240974 Chr.7: 31078102-31078102 on Build GRCh38
rs752786754 Chr.7: 31078136-31078136 on Build GRCh38
rs7804302 Chr.7: 31078031-31078031 on Build GRCh38
rs376849591 Chr.7: 31078091-31078091 on Build GRCh38
rs983706162 Chr.7: 31078109-31078109 on Build GRCh38
rs758341880 Chr.7: 31078138-31078138 on Build GRCh38
rs754777696 Chr.7: 31078108-31078108 on Build GRCh38
rs115677449 Chr.7: 31078168-31078168 on Build GRCh38
rs777331243 Chr.7: 31078034-31078034 on Build GRCh38
rs760771425 Chr.7: 31078099-31078099 on Build GRCh38
rs774764898 Chr.7: 31078078-31078078 on Build GRCh38
rs1025125460 Chr.7: 31078095-31078095 on Build GRCh38
rs756395323 Chr.7: 31077986-31077986 on Build GRCh38
rs371697914 Chr.7: 31078059-31078059 on Build GRCh38
rs754064970 Chr.7: 31078106-31078106 on Build GRCh38
rs1012265755 Chr.7: 31078064-31078064 on Build GRCh38
rs141094157 Chr.7: 31078003-31078003 on Build GRCh38
rs773440839 Chr.7: 31078094-31078094 on Build GRCh38

View Full Product Details