The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308097.1 | NP_001295026.1 | 2 - 3 | 252 | 150 |
NM_017769.4 | NP_060239.2 | 3 - 4 | 293 | 150 | |
XM_011536925.2 | XP_011535227.1 | 3 - 4 | 259 | 150 | |
XM_011536926.1 | XP_011535228.1 | 3 - 4 | 293 | 150 | |
GenBank mRNA | AB037754.1 | - | 3 - 4 | 245 | 150 |
AK000340.1 | - | 3 - 4 | 258 | 150 | |
AK093536.1 | - | 2 - 3 | 217 | 150 | |
AK225346.1 | - | 3 - 4 | 258 | 150 | |
AK295571.1 | - | 2 - 3 | 217 | 150 | |
AK296872.1 | - | 2 - 3 | 217 | 150 | |
BC000973.2 | - | 3 - 4 | 239 | 150 | |
DA491192.1 | - | 3 - 4 | 293 | 150 |