Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
Chromosome Location
Chr.8: 27733316 - 27772656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304529.1 NP_001291458.1 2 - 3 194 192
NM_001304530.1 NP_001291459.1 2 - 3 194 140
NM_001304532.1 NP_001291461.1 2 - 3 194 140
NM_018246.2 NP_060716.2 2 - 3 194 192
NR_130761.1 2 - 3 194 192
NR_130762.1 2 - 3 194 140
GenBank mRNA AK001715.1 - 2 - 3 194 140
AK027900.1 - 2 - 3 146 192
AK292875.1 - 2 - 3 181 140
AK294848.1 - 2 - 3 181 192
AK311437.1 - 2 - 3 157 140
AK316015.1 - 2 - 3 148 192
BC032588.1 - 2 - 3 183 140
BC049197.1 - 2 - 3 148 192
BC054039.1 - 2 - 3 134 192
BC065295.1 - 2 - 3 135 192
BC110391.1 - 2 - 3 135 140
BG718771.1 - 2 - 3 186 140
BU602778.1 - 2 - 3 195 192
DA218015.1 - 2 - 3 194 192

View Full Product Details