Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
Chromosome Location
Chr.8: 27733316 - 27772656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304532.1 NP_001291461.1 6 - 7 605 158
NM_018246.2 NP_060716.2 7 - 8 657 158
NR_130762.1 5 - 6 501 158
XM_011544570.2 XP_011542872.1 6 - 7 604 158
XM_011544571.2 XP_011542873.1 5 - 6 483 158
GenBank mRNA AK001715.1 - 6 - 7 605 158
AK027900.1 - 7 - 8 609 158
AK292875.1 - 6 - 7 592 158
AK294848.1 - 6 - 7 540 158
AK311437.1 - 5 - 6 464 158
BC032588.1 - 6 - 7 594 158
BC049197.1 - 7 - 8 611 158
BC065295.1 - 7 - 8 598 158
BC110391.1 - 5 - 6 442 158

View Full Product Details