Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10485385-10485652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986496 c.932G>A p.R311H Chr.18: 10485619-10485619 on Build GRCh38
986495 c.913G>A p.E305K Chr.18: 10485600-10485600 on Build GRCh38
708460 c.861G>C p.L287L Chr.18: 10485548-10485548 on Build GRCh38
2883173 c.822C>T p.D274D Chr.18: 10485509-10485509 on Build GRCh38
5001222 c.877G>T p.G293W Chr.18: 10485564-10485564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755780439 Chr.18: 10485479-10485479 on Build GRCh38
rs369783198 Chr.18: 10485467-10485467 on Build GRCh38
rs765925626 Chr.18: 10485610-10485610 on Build GRCh38
rs771067572 Chr.18: 10485501-10485501 on Build GRCh38
rs779292945 Chr.18: 10485555-10485555 on Build GRCh38
rs950729159 Chr.18: 10485408-10485408 on Build GRCh38
rs369608898 Chr.18: 10485509-10485509 on Build GRCh38
rs749914601 Chr.18: 10485533-10485533 on Build GRCh38
rs200699860 Chr.18: 10485628-10485628 on Build GRCh38
rs371900450 Chr.18: 10485500-10485500 on Build GRCh38
rs146444390 Chr.18: 10485480-10485480 on Build GRCh38
rs529396289 Chr.18: 10485513-10485513 on Build GRCh38
rs773464443 Chr.18: 10485424-10485424 on Build GRCh38
rs140388365 Chr.18: 10485563-10485563 on Build GRCh38
rs61741632 Chr.18: 10485506-10485506 on Build GRCh38
rs765577645 Chr.18: 10485617-10485617 on Build GRCh38
rs764129316 Chr.18: 10485448-10485448 on Build GRCh38
rs748706059 Chr.18: 10485494-10485494 on Build GRCh38
rs375344712 Chr.18: 10485412-10485412 on Build GRCh38
rs114821361 Chr.18: 10485552-10485552 on Build GRCh38
rs372734445 Chr.18: 10485554-10485554 on Build GRCh38
rs199818137 Chr.18: 10485615-10485615 on Build GRCh38
rs201834801 Chr.18: 10485583-10485583 on Build GRCh38
rs764648724 Chr.18: 10485441-10485441 on Build GRCh38
rs1015058758 Chr.18: 10485580-10485580 on Build GRCh38
rs779421245 Chr.18: 10485490-10485490 on Build GRCh38
rs772163651 Chr.18: 10485497-10485497 on Build GRCh38
rs765073166 Chr.18: 10485430-10485430 on Build GRCh38
rs936156933 Chr.18: 10485474-10485474 on Build GRCh38
rs144035486 Chr.18: 10485472-10485472 on Build GRCh38
rs917221215 Chr.18: 10485423-10485423 on Build GRCh38
rs528698632 Chr.18: 10485483-10485483 on Build GRCh38
rs772739500 Chr.18: 10485602-10485602 on Build GRCh38
rs374914282 Chr.18: 10485502-10485502 on Build GRCh38
rs991795534 Chr.18: 10485465-10485465 on Build GRCh38
rs958631444 Chr.18: 10485462-10485462 on Build GRCh38
rs750059738 Chr.18: 10485473-10485473 on Build GRCh38
rs771909445 Chr.18: 10485600-10485600 on Build GRCh38
rs760127834 Chr.18: 10485603-10485603 on Build GRCh38
rs747061093 Chr.18: 10485565-10485565 on Build GRCh38
rs755440394 Chr.18: 10485489-10485489 on Build GRCh38
rs760850682 Chr.18: 10485426-10485426 on Build GRCh38
rs746141096 Chr.18: 10485594-10485594 on Build GRCh38
rs150205350 Chr.18: 10485564-10485564 on Build GRCh38
rs762923907 Chr.18: 10485446-10485446 on Build GRCh38
rs753461711 Chr.18: 10485613-10485613 on Build GRCh38
rs753012867 Chr.18: 10485618-10485618 on Build GRCh38
rs751072365 Chr.18: 10485452-10485452 on Build GRCh38
rs369440825 Chr.18: 10485599-10485599 on Build GRCh38
rs539268005 Chr.18: 10485510-10485510 on Build GRCh38
rs377655181 Chr.18: 10485557-10485557 on Build GRCh38
rs1002886635 Chr.18: 10485598-10485598 on Build GRCh38
rs547035524 Chr.18: 10485499-10485499 on Build GRCh38
rs766984901 Chr.18: 10485468-10485468 on Build GRCh38
rs374082304 Chr.18: 10485595-10485595 on Build GRCh38
rs537305657 Chr.18: 10485591-10485591 on Build GRCh38
rs143107759 Chr.18: 10485550-10485550 on Build GRCh38
rs772140289 Chr.18: 10485414-10485414 on Build GRCh38
rs760308864 Chr.18: 10485545-10485545 on Build GRCh38
rs762869079 Chr.18: 10485505-10485505 on Build GRCh38
rs758870123 Chr.18: 10485619-10485619 on Build GRCh38

View Full Product Details