Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA3
Entrez Gene ID
3200
Gene Name
homeobox A3
Gene Aliases
HOX1, HOX1E
Chromosome Location
Chr.7: 27106190 - 27152581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030661.4 NP_109377.1 1 - 2 85 98
GenBank mRNA BC015180.1 - 1 - 2 85 98

View Full Product Details