The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242366.2 | NP_001229295.1 | 2 - 3 | 1176 | 68 |
GenBank mRNA | BC010691.1 | - | 2 - 3 | 1165 | 68 |