Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41907639-41907908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf51
Entrez Gene ID
285636
Gene Name
chromosome 5 open reading frame 51
Gene Aliases
-
UniGene
Hs.29899
Gene Chromosome Location
Chr.5: 41904313-41921636 on Build GRCh38
Transcripts
NM_175921.5 XM_005248289.4 XM_011514032.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1173136 c.187C>T p.L63F Chr.5: 41907822-41907822 on Build GRCh38
7197551 c.150G>T p.L50L Chr.5: 41907785-41907785 on Build GRCh38
5922432 c.179C>T p.P60L Chr.5: 41907814-41907814 on Build GRCh38
6616339 c.114T>C p.I38I Chr.5: 41907749-41907749 on Build GRCh38
5440098 c.132A>G p.L44L Chr.5: 41907767-41907767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372258134 Chr.5: 41907703-41907703 on Build GRCh38
rs888509685 Chr.5: 41907792-41907792 on Build GRCh38
rs556467579 Chr.5: 41907808-41907808 on Build GRCh38
rs747357874 Chr.5: 41907764-41907764 on Build GRCh38
rs866806591 Chr.5: 41907804-41907804 on Build GRCh38
rs367691957 Chr.5: 41907694-41907694 on Build GRCh38
rs755003533 Chr.5: 41907736-41907736 on Build GRCh38
rs773842836 Chr.5: 41907805-41907805 on Build GRCh38
rs148674200 Chr.5: 41907838-41907838 on Build GRCh38
rs190694566 Chr.5: 41907817-41907817 on Build GRCh38
rs753790572 Chr.5: 41907855-41907855 on Build GRCh38
rs575851208 Chr.5: 41907687-41907687 on Build GRCh38
rs1016476221 Chr.5: 41907864-41907864 on Build GRCh38
rs766365902 Chr.5: 41907826-41907826 on Build GRCh38
rs777273624 Chr.5: 41907782-41907782 on Build GRCh38
rs144983922 Chr.5: 41907751-41907751 on Build GRCh38
rs1002780560 Chr.5: 41907787-41907787 on Build GRCh38
rs372724399 Chr.5: 41907732-41907732 on Build GRCh38
rs779038385 Chr.5: 41907854-41907855 on Build GRCh38
rs1041369552 Chr.5: 41907737-41907737 on Build GRCh38
rs775761436 Chr.5: 41907694-41907694 on Build GRCh38
rs774192527 Chr.5: 41907837-41907837 on Build GRCh38
rs754880896 Chr.5: 41907857-41907857 on Build GRCh38
rs765159837 Chr.5: 41907690-41907690 on Build GRCh38
rs750176190 Chr.5: 41907705-41907705 on Build GRCh38
rs779940854 Chr.5: 41907723-41907723 on Build GRCh38
rs375771980 Chr.5: 41907733-41907733 on Build GRCh38
rs763636927 Chr.5: 41907819-41907819 on Build GRCh38
rs748326364 Chr.5: 41907750-41907750 on Build GRCh38
rs767106036 Chr.5: 41907815-41907815 on Build GRCh38
rs181848044 Chr.5: 41907825-41907825 on Build GRCh38
rs917065197 Chr.5: 41907720-41907720 on Build GRCh38
rs773737128 Chr.5: 41907752-41907752 on Build GRCh38
rs749405623 Chr.5: 41907725-41907725 on Build GRCh38
rs750608289 Chr.5: 41907867-41907867 on Build GRCh38
rs763525374 Chr.5: 41907801-41907801 on Build GRCh38
rs902855649 Chr.5: 41907747-41907747 on Build GRCh38
rs561142882 Chr.5: 41907749-41907749 on Build GRCh38
rs771607773 Chr.5: 41907781-41907781 on Build GRCh38
rs752940744 Chr.5: 41907785-41907785 on Build GRCh38
rs762581412 Chr.5: 41907797-41907797 on Build GRCh38
rs10061961 Chr.5: 41907707-41907707 on Build GRCh38

View Full Product Details