Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130110477-130110750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLP2
Entrez Gene ID
334
Gene Name
amyloid beta precursor like protein 2
Gene Aliases
APLP-2, APPH, APPL2, CDEBP
UniGene
Hs.370247
Gene Chromosome Location
Chr.11: 130069821-130144811 on Build GRCh38
Transcripts
NM_001142276.1 NM_001142277.1 NM_001142278.1 NM_001243299.1 NM_001642.2 NR_024515.1 NR_024516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4761304 c.376C>A p.R126S Chr.11: 130110634-130110634 on Build GRCh38
3729544 c.297G>T p.Q99H Chr.11: 130110555-130110555 on Build GRCh38
4761305 c.379T>A p.F127I Chr.11: 130110637-130110637 on Build GRCh38
6602218 c.363G>A p.K121K Chr.11: 130110621-130110621 on Build GRCh38
4973061 c.291G>A p.E97E Chr.11: 130110549-130110549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771968991 Chr.11: 130110523-130110523 on Build GRCh38
rs770017443 Chr.11: 130110538-130110538 on Build GRCh38
rs773428429 Chr.11: 130110540-130110540 on Build GRCh38
rs142512603 Chr.11: 130110583-130110583 on Build GRCh38
rs776469245 Chr.11: 130110651-130110651 on Build GRCh38
rs147726545 Chr.11: 130110661-130110661 on Build GRCh38
rs746803432 Chr.11: 130110524-130110524 on Build GRCh38
rs774396553 Chr.11: 130110654-130110654 on Build GRCh38
rs567698773 Chr.11: 130110511-130110512 on Build GRCh38
rs759561514 Chr.11: 130110657-130110657 on Build GRCh38
rs770041870 Chr.11: 130110672-130110673 on Build GRCh38
rs762963531 Chr.11: 130110558-130110558 on Build GRCh38
rs201076312 Chr.11: 130110671-130110671 on Build GRCh38
rs756829089 Chr.11: 130110505-130110505 on Build GRCh38
rs756462049 Chr.11: 130110607-130110607 on Build GRCh38
rs150893992 Chr.11: 130110626-130110626 on Build GRCh38
rs150028786 Chr.11: 130110660-130110660 on Build GRCh38
rs369206839 Chr.11: 130110585-130110585 on Build GRCh38
rs376766945 Chr.11: 130110502-130110502 on Build GRCh38
rs979851900 Chr.11: 130110721-130110721 on Build GRCh38
rs765031023 Chr.11: 130110690-130110690 on Build GRCh38
rs199647187 Chr.11: 130110637-130110637 on Build GRCh38
rs764538127 Chr.11: 130110675-130110675 on Build GRCh38
rs866931273 Chr.11: 130110624-130110624 on Build GRCh38
rs763769514 Chr.11: 130110608-130110608 on Build GRCh38
rs749909473 Chr.11: 130110693-130110693 on Build GRCh38
rs531144318 Chr.11: 130110629-130110629 on Build GRCh38
rs756937279 Chr.11: 130110685-130110685 on Build GRCh38
rs762067675 Chr.11: 130110527-130110530 on Build GRCh38
rs758333669 Chr.11: 130110635-130110635 on Build GRCh38
rs568147308 Chr.11: 130110584-130110584 on Build GRCh38
rs748764006 Chr.11: 130110499-130110499 on Build GRCh38
rs753551329 Chr.11: 130110610-130110610 on Build GRCh38
rs761038664 Chr.11: 130110669-130110669 on Build GRCh38
rs373959494 Chr.11: 130110618-130110618 on Build GRCh38
rs767962387 Chr.11: 130110586-130110586 on Build GRCh38
rs745785358 Chr.11: 130110512-130110512 on Build GRCh38
rs376319356 Chr.11: 130110679-130110679 on Build GRCh38
rs553545609 Chr.11: 130110590-130110590 on Build GRCh38
rs144808175 Chr.11: 130110509-130110509 on Build GRCh38
rs768351062 Chr.11: 130110644-130110644 on Build GRCh38
rs368311631 Chr.11: 130110588-130110588 on Build GRCh38
rs138645940 Chr.11: 130110653-130110653 on Build GRCh38

View Full Product Details