Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 111079313-111079564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA2A
Entrez Gene ID
150
Gene Name
adrenoceptor alpha 2A
Gene Aliases
ADRA2, ADRA2R, ADRAR, ALPHA2AAR, ZNF32
UniGene
Hs.249159
Gene Chromosome Location
Chr.10: 111077032-111080907 on Build GRCh38
Transcripts
NM_000681.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415169 c.1323C>T p.L441L Chr.10: 111079364-111079364 on Build GRCh38
4526119 c.1328G>T p.R443L Chr.10: 111079369-111079369 on Build GRCh38
7323760 c.1287C>T p.F429F Chr.10: 111079328-111079328 on Build GRCh38
2052961 c.1305C>T p.R435R Chr.10: 111079346-111079346 on Build GRCh38
6595500 c.1306G>A p.A436T Chr.10: 111079347-111079347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs33931318 Chr.10: 111079413-111079413 on Build GRCh38
rs150599677 Chr.10: 111079534-111079534 on Build GRCh38
rs778105791 Chr.10: 111079368-111079369 on Build GRCh38
rs547631131 Chr.10: 111079372-111079372 on Build GRCh38
rs753064524 Chr.10: 111079354-111079354 on Build GRCh38
rs913909924 Chr.10: 111079455-111079455 on Build GRCh38
rs764891418 Chr.10: 111079347-111079347 on Build GRCh38
rs369219397 Chr.10: 111079369-111079369 on Build GRCh38
rs997668339 Chr.10: 111079536-111079536 on Build GRCh38
rs926757757 Chr.10: 111079424-111079424 on Build GRCh38
rs771778912 Chr.10: 111079396-111079396 on Build GRCh38
rs200026350 Chr.10: 111079436-111079436 on Build GRCh38
rs776154544 Chr.10: 111079343-111079343 on Build GRCh38
rs765866331 Chr.10: 111079342-111079342 on Build GRCh38
rs369765179 Chr.10: 111079482-111079482 on Build GRCh38
rs111623406 Chr.10: 111079433-111079433 on Build GRCh38
rs1006150720 Chr.10: 111079390-111079390 on Build GRCh38
rs950647547 Chr.10: 111079374-111079374 on Build GRCh38
rs762388343 Chr.10: 111079445-111079445 on Build GRCh38
rs375358019 Chr.10: 111079438-111079438 on Build GRCh38
rs943577558 Chr.10: 111079512-111079512 on Build GRCh38
rs371747133 Chr.10: 111079502-111079502 on Build GRCh38
rs562832369 Chr.10: 111079453-111079453 on Build GRCh38
rs756360648 Chr.10: 111079383-111079383 on Build GRCh38
rs764363447 Chr.10: 111079439-111079439 on Build GRCh38
rs759161719 Chr.10: 111079345-111079345 on Build GRCh38
rs551131283 Chr.10: 111079456-111079456 on Build GRCh38
rs751998895 Chr.10: 111079366-111079366 on Build GRCh38
rs760720691 Chr.10: 111079337-111079337 on Build GRCh38
rs764602189 Chr.10: 111079364-111079364 on Build GRCh38
rs747913823 Chr.10: 111079389-111079389 on Build GRCh38
rs533089129 Chr.10: 111079454-111079454 on Build GRCh38
rs751995817 Chr.10: 111079440-111079440 on Build GRCh38
rs770901159 Chr.10: 111079412-111079412 on Build GRCh38
rs750581149 Chr.10: 111079370-111079370 on Build GRCh38
rs1043973146 Chr.10: 111079484-111079484 on Build GRCh38
rs939555133 Chr.10: 111079463-111079463 on Build GRCh38
rs980316108 Chr.10: 111079419-111079419 on Build GRCh38
rs746887236 Chr.10: 111079411-111079411 on Build GRCh38
rs776502468 Chr.10: 111079509-111079509 on Build GRCh38
rs759077809 Chr.10: 111079421-111079421 on Build GRCh38
rs763230238 Chr.10: 111079360-111079360 on Build GRCh38
rs772013593 Chr.10: 111079368-111079368 on Build GRCh38
rs780174433 Chr.10: 111079384-111079384 on Build GRCh38
rs1039592811 Chr.10: 111079516-111079516 on Build GRCh38
rs747221093 Chr.10: 111079439-111079440 on Build GRCh38
rs34303217 Chr.10: 111079524-111079524 on Build GRCh38
rs1047479759 Chr.10: 111079541-111079541 on Build GRCh38

View Full Product Details