Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70735138-70735383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
-
UniGene
Hs.352156
Gene Chromosome Location
Chr.10: 70672518-70762441 on Build GRCh38
Transcripts
NM_080722.4 NM_139155.3 XM_011539300.2 XM_011539301.2 XM_011539302.2 XM_011539303.2 XM_011539305.2 XM_011539306.2 XM_011539307.2 XM_011539308.2 XM_011539309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4659476 c.1456C>T p.R486C Chr.10: 70735263-70735263 on Build GRCh38
2152669 c.1476C>T p.G492G Chr.10: 70735283-70735283 on Build GRCh38
308897 c.1428G>T p.G476G Chr.10: 70735235-70735235 on Build GRCh38
6130017 c.1480C>G p.Q494E Chr.10: 70735287-70735287 on Build GRCh38
3807598 c.1457G>A p.R486H Chr.10: 70735264-70735264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765242015 Chr.10: 70735222-70735222 on Build GRCh38
rs564600362 Chr.10: 70735216-70735216 on Build GRCh38
rs780215561 Chr.10: 70735229-70735229 on Build GRCh38
rs140097899 Chr.10: 70735348-70735348 on Build GRCh38
rs781417210 Chr.10: 70735264-70735264 on Build GRCh38
rs373868260 Chr.10: 70735157-70735157 on Build GRCh38
rs749627609 Chr.10: 70735291-70735291 on Build GRCh38
rs775161249 Chr.10: 70735316-70735316 on Build GRCh38
rs767240397 Chr.10: 70735213-70735213 on Build GRCh38
rs373853037 Chr.10: 70735188-70735188 on Build GRCh38
rs747276945 Chr.10: 70735258-70735258 on Build GRCh38
rs148351062 Chr.10: 70735256-70735256 on Build GRCh38
rs367637583 Chr.10: 70735175-70735175 on Build GRCh38
rs767274604 Chr.10: 70735315-70735315 on Build GRCh38
rs201655502 Chr.10: 70735323-70735323 on Build GRCh38
rs762614707 Chr.10: 70735193-70735193 on Build GRCh38
rs760576349 Chr.10: 70735327-70735327 on Build GRCh38
rs755319194 Chr.10: 70735263-70735263 on Build GRCh38
rs780137633 Chr.10: 70735223-70735223 on Build GRCh38
rs1004163289 Chr.10: 70735184-70735184 on Build GRCh38
rs769284473 Chr.10: 70735189-70735189 on Build GRCh38
rs1041357648 Chr.10: 70735342-70735342 on Build GRCh38
rs928634234 Chr.10: 70735192-70735192 on Build GRCh38
rs1048606353 Chr.10: 70735319-70735319 on Build GRCh38
rs759144393 Chr.10: 70735314-70735314 on Build GRCh38
rs750734959 Chr.10: 70735171-70735171 on Build GRCh38
rs756007850 Chr.10: 70735217-70735217 on Build GRCh38
rs546518440 Chr.10: 70735209-70735209 on Build GRCh38
rs143371154 Chr.10: 70735187-70735187 on Build GRCh38
rs774188465 Chr.10: 70735204-70735204 on Build GRCh38
rs117467557 Chr.10: 70735165-70735165 on Build GRCh38
rs772865695 Chr.10: 70735190-70735190 on Build GRCh38
rs774098828 Chr.10: 70735304-70735304 on Build GRCh38
rs747364881 Chr.10: 70735169-70735169 on Build GRCh38
rs766124715 Chr.10: 70735195-70735195 on Build GRCh38
rs918701359 Chr.10: 70735212-70735212 on Build GRCh38
rs375859238 Chr.10: 70735338-70735338 on Build GRCh38
rs973225992 Chr.10: 70735202-70735202 on Build GRCh38
rs201013821 Chr.10: 70735339-70735339 on Build GRCh38
rs1029575652 Chr.10: 70735232-70735232 on Build GRCh38
rs777171630 Chr.10: 70735176-70735176 on Build GRCh38
rs773002259 Chr.10: 70735326-70735327 on Build GRCh38
rs748634034 Chr.10: 70735265-70735265 on Build GRCh38
rs200090127 Chr.10: 70735294-70735294 on Build GRCh38
rs112010223 Chr.10: 70735260-70735260 on Build GRCh38
rs773927392 Chr.10: 70735289-70735289 on Build GRCh38
rs770192483 Chr.10: 70735286-70735286 on Build GRCh38
rs758535322 Chr.10: 70735224-70735224 on Build GRCh38

View Full Product Details