Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7091205-7091468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
UniGene
Hs.397705
Gene Chromosome Location
Chr.1: 6785324-7769706 on Build GRCh38
Transcripts
NM_015215.3 XM_011541083.2 XM_011541084.2 XM_011541085.2 XM_011541086.2 XM_011541087.2 XM_011541088.2 XM_011541089.2 XM_011541090.2 XM_011541091.2 XM_011541092.2 XM_017000773.1 XM_017000774.1 XM_017000775.1 XM_017000776.1 XM_017000777.1 XM_017000778.1 XM_017000779.1 XM_017000780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211493 c.255A>T p.I85I Chr.1: 7091324-7091324 on Build GRCh38
3492177 c.269A>G p.H90R Chr.1: 7091338-7091338 on Build GRCh38
6211492 c.255A>T p.I85I Chr.1: 7091324-7091324 on Build GRCh38
6286634 c.273A>T p.E91D Chr.1: 7091342-7091342 on Build GRCh38
3866062 c.274G>A p.E92K Chr.1: 7091343-7091343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779451928 Chr.1: 7091336-7091336 on Build GRCh38
rs762589698 Chr.1: 7091414-7091414 on Build GRCh38
rs531861098 Chr.1: 7091407-7091407 on Build GRCh38
rs896170631 Chr.1: 7091251-7091251 on Build GRCh38
rs970863807 Chr.1: 7091250-7091250 on Build GRCh38
rs1023544414 Chr.1: 7091273-7091273 on Build GRCh38
rs772486063 Chr.1: 7091345-7091345 on Build GRCh38
rs780132556 Chr.1: 7091280-7091280 on Build GRCh38
rs749758954 Chr.1: 7091309-7091309 on Build GRCh38
rs146654528 Chr.1: 7091339-7091339 on Build GRCh38
rs756093964 Chr.1: 7091265-7091265 on Build GRCh38
rs45517936 Chr.1: 7091445-7091445 on Build GRCh38
rs1012113590 Chr.1: 7091253-7091253 on Build GRCh38
rs879253761 Chr.1: 7091337-7091337 on Build GRCh38
rs560909558 Chr.1: 7091405-7091405 on Build GRCh38
rs776248421 Chr.1: 7091386-7091386 on Build GRCh38
rs772681264 Chr.1: 7091356-7091356 on Build GRCh38
rs72640073 Chr.1: 7091419-7091419 on Build GRCh38
rs746696309 Chr.1: 7091359-7091359 on Build GRCh38
rs967814138 Chr.1: 7091282-7091282 on Build GRCh38
rs767630728 Chr.1: 7091257-7091257 on Build GRCh38
rs1034674536 Chr.1: 7091248-7091248 on Build GRCh38
rs755452751 Chr.1: 7091315-7091315 on Build GRCh38
rs775564254 Chr.1: 7091410-7091410 on Build GRCh38
rs865887247 Chr.1: 7091376-7091376 on Build GRCh38
rs200492972 Chr.1: 7091259-7091259 on Build GRCh38
rs770522792 Chr.1: 7091381-7091381 on Build GRCh38
rs868101025 Chr.1: 7091311-7091311 on Build GRCh38
rs982300779 Chr.1: 7091261-7091261 on Build GRCh38

View Full Product Details