Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 153485926-153486191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf79
Entrez Gene ID
152118
Gene Name
chromosome 3 open reading frame 79
Gene Aliases
-
UniGene
Hs.58586
Gene Chromosome Location
Chr.3: 153484495-153502697 on Build GRCh38
Transcripts
NM_001101337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5437484 c.159A>G p.P53P Chr.3: 153486041-153486041 on Build GRCh38
398221 c.157C>G p.P53A Chr.3: 153486039-153486039 on Build GRCh38
1040109 c.201T>G p.I67M Chr.3: 153486083-153486083 on Build GRCh38
7200831 c.204G>A p.L68L Chr.3: 153486086-153486086 on Build GRCh38
5160513 c.199_200insT p.L68fs*16 Chr.3: 153486081-153486082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749672468 Chr.3: 153485992-153485992 on Build GRCh38
rs181728536 Chr.3: 153485989-153485989 on Build GRCh38
rs375129889 Chr.3: 153486009-153486009 on Build GRCh38
rs754910353 Chr.3: 153485992-153485992 on Build GRCh38
rs200484423 Chr.3: 153486055-153486055 on Build GRCh38
rs1049394647 Chr.3: 153486156-153486156 on Build GRCh38
rs780047914 Chr.3: 153485980-153485980 on Build GRCh38
rs1000960323 Chr.3: 153486063-153486063 on Build GRCh38
rs575130185 Chr.3: 153485970-153485970 on Build GRCh38
rs754095439 Chr.3: 153486078-153486078 on Build GRCh38
rs889472788 Chr.3: 153486158-153486158 on Build GRCh38
rs777313562 Chr.3: 153486151-153486151 on Build GRCh38
rs769373844 Chr.3: 153486006-153486006 on Build GRCh38
rs774619926 Chr.3: 153486037-153486037 on Build GRCh38
rs757598822 Chr.3: 153486100-153486100 on Build GRCh38
rs751339794 Chr.3: 153486124-153486124 on Build GRCh38
rs750174814 Chr.3: 153485966-153485966 on Build GRCh38
rs1039963245 Chr.3: 153486024-153486024 on Build GRCh38
rs773715693 Chr.3: 153485991-153485992 on Build GRCh38
rs1032462449 Chr.3: 153486075-153486075 on Build GRCh38
rs543244911 Chr.3: 153486076-153486076 on Build GRCh38
rs57748412 Chr.3: 153486020-153486020 on Build GRCh38
rs771264913 Chr.3: 153486021-153486021 on Build GRCh38
rs374950907 Chr.3: 153486125-153486125 on Build GRCh38
rs780999258 Chr.3: 153486131-153486131 on Build GRCh38
rs1009594420 Chr.3: 153486106-153486106 on Build GRCh38
rs372708596 Chr.3: 153486061-153486061 on Build GRCh38
rs748908605 Chr.3: 153486154-153486154 on Build GRCh38
rs760049407 Chr.3: 153486050-153486050 on Build GRCh38
rs867024892 Chr.3: 153485991-153485991 on Build GRCh38
rs770294033 Chr.3: 153485991-153485991 on Build GRCh38
rs981343989 Chr.3: 153486013-153486013 on Build GRCh38
rs775580511 Chr.3: 153486053-153486053 on Build GRCh38
rs931058560 Chr.3: 153486145-153486145 on Build GRCh38
rs184466277 Chr.3: 153486141-153486141 on Build GRCh38
rs56196219 Chr.3: 153486136-153486136 on Build GRCh38
rs201155059 Chr.3: 153486110-153486110 on Build GRCh38
rs1031787546 Chr.3: 153485967-153485967 on Build GRCh38
rs970985304 Chr.3: 153485954-153485954 on Build GRCh38
rs899629508 Chr.3: 153486059-153486059 on Build GRCh38
rs879252052 Chr.3: 153486123-153486123 on Build GRCh38
rs202116458 Chr.3: 153485995-153485995 on Build GRCh38

View Full Product Details