Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 214947357-214947622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA12
Entrez Gene ID
26154
Gene Name
ATP binding cassette subfamily A member 12
Gene Aliases
ARCI4A, ARCI4B, ICR2B, LI2
UniGene
Hs.134585
Gene Chromosome Location
Chr.2: 214931542-215138591 on Build GRCh38
Transcripts
NM_015657.3 NM_173076.2 NR_103740.1 XM_011510951.2 NR_110292.1

Gene Symbol
LOC101928103
Entrez Gene ID
101928103
Gene Name
uncharacterized LOC101928103
Gene Aliases
-
UniGene
Hs.665619
Gene Chromosome Location
Chr.2: 214810229-214963605 on Build GRCh38
Transcripts
NM_015657.3 NM_173076.2 NR_103740.1 XM_011510951.2 NR_110292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6293823 c.7136T>A p.L2379Q Chr.2: 214947525-214947525 on Build GRCh38
4441202 c.6269C>A p.P2090H Chr.2: 214947438-214947438 on Build GRCh38
6090178 c.7114A>G p.K2372E Chr.2: 214947547-214947547 on Build GRCh38
1181210 c.7207G>T p.A2403S Chr.2: 214947454-214947454 on Build GRCh38
4091009 c.7173T>C p.S2391S Chr.2: 214947488-214947488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990368624 Chr.2: 214947534-214947534 on Build GRCh38
rs747667327 Chr.2: 214947397-214947397 on Build GRCh38
rs2274412 Chr.2: 214947573-214947573 on Build GRCh38
rs758004412 Chr.2: 214947402-214947402 on Build GRCh38
rs781732646 Chr.2: 214947587-214947587 on Build GRCh38
rs957495468 Chr.2: 214947496-214947496 on Build GRCh38
rs752156383 Chr.2: 214947552-214947552 on Build GRCh38
rs372585292 Chr.2: 214947422-214947422 on Build GRCh38
rs754933913 Chr.2: 214947548-214947548 on Build GRCh38
rs977651634 Chr.2: 214947439-214947439 on Build GRCh38
rs368391238 Chr.2: 214947395-214947395 on Build GRCh38
rs764302965 Chr.2: 214947485-214947485 on Build GRCh38
rs760482242 Chr.2: 214947527-214947527 on Build GRCh38
rs771375124 Chr.2: 214947447-214947447 on Build GRCh38
rs746403011 Chr.2: 214947452-214947452 on Build GRCh38
rs774514604 Chr.2: 214947524-214947524 on Build GRCh38
rs762022735 Chr.2: 214947572-214947573 on Build GRCh38
rs775490909 Chr.2: 214947454-214947454 on Build GRCh38
rs777592577 Chr.2: 214947577-214947577 on Build GRCh38
rs770215233 Chr.2: 214947593-214947593 on Build GRCh38
rs966511460 Chr.2: 214947434-214947434 on Build GRCh38
rs747411339 Chr.2: 214947443-214947443 on Build GRCh38
rs754001328 Chr.2: 214947524-214947524 on Build GRCh38
rs777524736 Chr.2: 214947412-214947412 on Build GRCh38
rs754933102 Chr.2: 214947389-214947389 on Build GRCh38
rs763031250 Chr.2: 214947481-214947481 on Build GRCh38
rs756939478 Chr.2: 214947584-214947584 on Build GRCh38
rs746709997 Chr.2: 214947581-214947581 on Build GRCh38
rs753595337 Chr.2: 214947544-214947544 on Build GRCh38
rs752220062 Chr.2: 214947535-214947535 on Build GRCh38
rs765117594 Chr.2: 214947440-214947440 on Build GRCh38
rs894032185 Chr.2: 214947396-214947396 on Build GRCh38

View Full Product Details