Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159993376-159993621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
UniGene
Hs.673
Gene Chromosome Location
Chr.3: 159988836-159996019 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Gene Symbol
IL12A-AS1
Entrez Gene ID
101928376
Gene Name
IL12A antisense RNA 1
Gene Aliases
-
UniGene
Hs.565865
Gene Chromosome Location
Chr.3: 159913400-160207092 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832004 c.410C>G p.S137C Chr.3: 159993482-159993482 on Build GRCh38
6684831 c.387T>C p.S129S Chr.3: 159993459-159993459 on Build GRCh38
7200036 c.384G>T p.E128D Chr.3: 159993456-159993456 on Build GRCh38
5505830 c.396delT p.S133fs*7 Chr.3: 159993468-159993468 on Build GRCh38
4927930 c.401G>C p.R134T Chr.3: 159993473-159993473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980092801 Chr.3: 159993432-159993432 on Build GRCh38
rs751284391 Chr.3: 159993519-159993520 on Build GRCh38
rs761130017 Chr.3: 159993564-159993564 on Build GRCh38
rs776668796 Chr.3: 159993582-159993582 on Build GRCh38
rs200596291 Chr.3: 159993454-159993454 on Build GRCh38
rs866340019 Chr.3: 159993475-159993475 on Build GRCh38
rs923023325 Chr.3: 159993559-159993559 on Build GRCh38
rs779949936 Chr.3: 159993518-159993519 on Build GRCh38
rs745535302 Chr.3: 159993547-159993547 on Build GRCh38
rs768060361 Chr.3: 159993418-159993418 on Build GRCh38
rs775946182 Chr.3: 159993557-159993557 on Build GRCh38
rs764623851 Chr.3: 159993573-159993573 on Build GRCh38
rs765492561 Chr.3: 159993592-159993592 on Build GRCh38
rs761183293 Chr.3: 159993424-159993424 on Build GRCh38
rs139950534 Chr.3: 159993462-159993462 on Build GRCh38
rs753948534 Chr.3: 159993443-159993443 on Build GRCh38
rs778132771 Chr.3: 159993532-159993534 on Build GRCh38
rs913028578 Chr.3: 159993505-159993505 on Build GRCh38
rs9832255 Chr.3: 159993541-159993541 on Build GRCh38
rs748044888 Chr.3: 159993507-159993507 on Build GRCh38
rs758221067 Chr.3: 159993436-159993437 on Build GRCh38
rs753074278 Chr.3: 159993421-159993421 on Build GRCh38
rs1040208561 Chr.3: 159993549-159993549 on Build GRCh38
rs567430718 Chr.3: 159993449-159993449 on Build GRCh38
rs772263185 Chr.3: 159993552-159993552 on Build GRCh38
rs756393736 Chr.3: 159993433-159993433 on Build GRCh38
rs779055728 Chr.3: 159993470-159993470 on Build GRCh38
rs990104054 Chr.3: 159993497-159993497 on Build GRCh38
rs754797956 Chr.3: 159993487-159993487 on Build GRCh38
rs375505799 Chr.3: 159993519-159993519 on Build GRCh38
rs773938409 Chr.3: 159993536-159993536 on Build GRCh38
rs770692194 Chr.3: 159993525-159993525 on Build GRCh38
rs756428573 Chr.3: 159993526-159993527 on Build GRCh38
rs748850178 Chr.3: 159993522-159993522 on Build GRCh38
rs750544982 Chr.3: 159993594-159993594 on Build GRCh38
rs201655135 Chr.3: 159993478-159993478 on Build GRCh38
rs761941291 Chr.3: 159993586-159993586 on Build GRCh38
rs767700076 Chr.3: 159993431-159993431 on Build GRCh38
rs374624685 Chr.3: 159993501-159993501 on Build GRCh38
rs375527576 Chr.3: 159993509-159993509 on Build GRCh38

View Full Product Details