Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66638364-66638591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890215 c.448G>T p.E150* Chr.7: 66638386-66638386 on Build GRCh38
3882077 c.457C>T p.R153C Chr.7: 66638395-66638395 on Build GRCh38
3367006 c.490A>G p.K164E Chr.7: 66638428-66638428 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531332669 Chr.7: 66638551-66638551 on Build GRCh38
rs751667494 Chr.7: 66638463-66638463 on Build GRCh38
rs1006793740 Chr.7: 66638549-66638549 on Build GRCh38
rs397720861 Chr.7: 66638452-66638453 on Build GRCh38
rs781613111 Chr.7: 66638453-66638454 on Build GRCh38
rs938944327 Chr.7: 66638516-66638516 on Build GRCh38
rs989198210 Chr.7: 66638540-66638540 on Build GRCh38
rs886648324 Chr.7: 66638518-66638518 on Build GRCh38
rs765235486 Chr.7: 66638396-66638396 on Build GRCh38
rs182290415 Chr.7: 66638517-66638517 on Build GRCh38
rs755121011 Chr.7: 66638468-66638468 on Build GRCh38
rs978399798 Chr.7: 66638554-66638554 on Build GRCh38
rs762715865 Chr.7: 66638506-66638506 on Build GRCh38
rs886645555 Chr.7: 66638445-66638445 on Build GRCh38
rs756458562 Chr.7: 66638572-66638572 on Build GRCh38
rs57580125 Chr.7: 66638448-66638449 on Build GRCh38
rs915073290 Chr.7: 66638550-66638550 on Build GRCh38
rs761707159 Chr.7: 66638395-66638395 on Build GRCh38
rs796052686 Chr.7: 66638394-66638394 on Build GRCh38
rs906179618 Chr.7: 66638502-66638502 on Build GRCh38
rs191592577 Chr.7: 66638482-66638482 on Build GRCh38
rs187113750 Chr.7: 66638457-66638457 on Build GRCh38
rs540783571 Chr.7: 66638424-66638424 on Build GRCh38
rs767984903 Chr.7: 66638385-66638385 on Build GRCh38
rs776661654 Chr.7: 66638386-66638386 on Build GRCh38
rs368833598 Chr.7: 66638507-66638507 on Build GRCh38
rs879025829 Chr.7: 66638439-66638439 on Build GRCh38
rs74362924 Chr.7: 66638488-66638488 on Build GRCh38
rs773209661 Chr.7: 66638403-66638403 on Build GRCh38
rs546503242 Chr.7: 66638494-66638494 on Build GRCh38

View Full Product Details