Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219239055-219239328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L
Entrez Gene ID
79411
Gene Name
galactosidase beta 1 like
Gene Aliases
-
UniGene
Hs.181173
Gene Chromosome Location
Chr.2: 219236598-219245429 on Build GRCh38
Transcripts
NM_001286423.1 NM_001286427.1 NM_024506.4 XM_011511813.2 XM_017004895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4382832 c.1045C>T p.R349* Chr.2: 219239109-219239109 on Build GRCh38
719906 c.1009G>C p.E337Q Chr.2: 219239145-219239145 on Build GRCh38
1016655 c.1046G>A p.R349Q Chr.2: 219239108-219239108 on Build GRCh38
719907 c.1032G>A p.K344K Chr.2: 219239122-219239122 on Build GRCh38
3709423 c.1031A>C p.K344T Chr.2: 219239123-219239123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016724480 Chr.2: 219239090-219239090 on Build GRCh38
rs3215296 Chr.2: 219239301-219239301 on Build GRCh38
rs766106152 Chr.2: 219239109-219239109 on Build GRCh38
rs746089587 Chr.2: 219239220-219239220 on Build GRCh38
rs747346150 Chr.2: 219239151-219239151 on Build GRCh38
rs559126808 Chr.2: 219239111-219239111 on Build GRCh38
rs771195423 Chr.2: 219239159-219239159 on Build GRCh38
rs962107035 Chr.2: 219239100-219239100 on Build GRCh38
rs397840074 Chr.2: 219239305-219239305 on Build GRCh38
rs566204911 Chr.2: 219239182-219239182 on Build GRCh38
rs775949865 Chr.2: 219239229-219239229 on Build GRCh38
rs752018394 Chr.2: 219239247-219239247 on Build GRCh38
rs878922322 Chr.2: 219239123-219239123 on Build GRCh38
rs148493267 Chr.2: 219239193-219239193 on Build GRCh38
rs943547645 Chr.2: 219239274-219239274 on Build GRCh38
rs781420753 Chr.2: 219239162-219239162 on Build GRCh38
rs945092141 Chr.2: 219239266-219239266 on Build GRCh38
rs776594285 Chr.2: 219239227-219239230 on Build GRCh38
rs775415487 Chr.2: 219239167-219239167 on Build GRCh38
rs775490864 Chr.2: 219239289-219239289 on Build GRCh38
rs753262485 Chr.2: 219239121-219239121 on Build GRCh38
rs766238132 Chr.2: 219239191-219239191 on Build GRCh38
rs1037693229 Chr.2: 219239160-219239160 on Build GRCh38
rs754549759 Chr.2: 219239112-219239112 on Build GRCh38
rs147170651 Chr.2: 219239163-219239163 on Build GRCh38
rs778180679 Chr.2: 219239150-219239150 on Build GRCh38
rs778219795 Chr.2: 219239114-219239114 on Build GRCh38
rs761876425 Chr.2: 219239179-219239179 on Build GRCh38
rs757620758 Chr.2: 219239258-219239258 on Build GRCh38
rs750238214 Chr.2: 219239091-219239091 on Build GRCh38
rs761189432 Chr.2: 219239285-219239285 on Build GRCh38
rs889350570 Chr.2: 219239116-219239116 on Build GRCh38
rs772270225 Chr.2: 219239259-219239259 on Build GRCh38
rs573301219 Chr.2: 219239263-219239263 on Build GRCh38
rs760567051 Chr.2: 219239188-219239188 on Build GRCh38
rs758821715 Chr.2: 219239128-219239128 on Build GRCh38
rs942286368 Chr.2: 219239138-219239138 on Build GRCh38
rs972464906 Chr.2: 219239125-219239125 on Build GRCh38
rs142820300 Chr.2: 219239192-219239192 on Build GRCh38
rs755869238 Chr.2: 219239108-219239108 on Build GRCh38
rs752148143 Chr.2: 219239210-219239210 on Build GRCh38
rs778233676 Chr.2: 219239230-219239230 on Build GRCh38
rs767403391 Chr.2: 219239083-219239083 on Build GRCh38
rs779216284 Chr.2: 219239183-219239183 on Build GRCh38
rs554872974 Chr.2: 219239204-219239204 on Build GRCh38

View Full Product Details